The ProSightPD application generates a results report that contains a list of the matching proteoforms identified by the search nodes that you selected, as shown in the following figure. You can access the results report through the application’s Start page or File menu. Through the Job Queue, you can access the results, including a failed workflow analysis. The application can save multiple search results within a single report.
Figure Results reports displayed in the application window
12345
No.DescriptionNo.Description
1
Result Table Chooser
4
Results table
2
List of open pages
5
Field Chooser
3
Status bar