The Proteoform and Protein Filter node filters proteoforms based on confidence.
The following table lists the node parameters.
Parameter | Description |
|---|---|
1. Proteoform Filters | |
Proteoform Confidence at Least | Proteoforms with a lower confidence than specified are excluded from the final result:
You can define confidence levels in the configuration panel of the Administration page. |
Keep Lower Confidence PrSMs | If set to True, keep PrSMs with lower confidence than specified at the proteoform level. If set to False, do not keep them. (Default) |