The following topics describe the nodes that search the assigned database.
- Annotated Proteoform Search node
- Single Proteoform Search node
- Subsequence Search node
- Tag to Annotated Proteoform Search node
- Tag to Subsequence Search
NOTE
The ProSightPD 4.4 application includes the legacy search nodes from ProSightPD 3.0 so that you can reproduce previous results in the ProSightPD 4.4 application. These legacy nodes require a database in the .pscw file format generated in ProSightPC. You cannot directly convert .pscw to .psdb.
Use the source .xml file, which the .pscw file was generated from, to generate a .psdb-formatted database. To use a legacy search node, replace the 4.4 node with the equivalent legacy node and review all parameters to ensure agreement between your previous ProSightPD 3.0 workflows and your ProSightPD 4.4 workflows.