The following table contains known defects in the software, categorized by software section, with a brief abstract and information related to each defect’s severity and risk. Product management assesses risk, which can differ significantly from the reported severity.
Software section | Severity | Abstract | Risk |
---|---|---|---|
Workflow management | Medium | FDR cannot be calculated for workflows containing both Med/High and Hi/Hi data types. Split into two workflows. | Low |
Fragment Match View | Medium | For larger proteoforms the fragment match view may load slowly. | Medium |
No support for ETciD activation | Minor | Override auto and select EThcD. | Minor |