This release incorporates these new or enhanced features:
- Post-translational modification site ambiguity display in the ProSightPD Fragment Map and PrSMs and Proteoform tables
- Random Seed Value parameter added to most search nodes to ensure FDR calculation reproducibility