Jump to main content
The ProSight Native application does not automatically save your project. Use the Save button to save your project for easy access in the future.
When all the correct settings and results are completed for the data set, all the information can be saved as a ProSight Native project.
Save a ProSight Native project
Procedure
-
To edit a project name, select the pencil icon next to the Project Name and type in a descriptive name.
Note: The project name will not affect the file name for the project.
- Select the checkmark icon to confirm the new project name.
-
Select the Save File icon in the upper-left corner.
The entered project name is autofilled as the File Name of the .psn file.
- (Optional) Enter a different File Name for the .psn file, if desired.
- Select Save.
-
Select the Home icon in the upper-left corner to return to the Home screen.
The new project appears in the Recent Projects column of the Home screen.