Jump to main content
The Top-Down Search workflow is designed for deconvolution and identification of proteoform masses using the top-down search engine.
The Top-Down Search workflow
The Top-Down Search workflow contains the TD Search #1 and Results tabs.
Create a Top-Down Search workflow
Procedure
- From the Home screen, select the Top-Down Search workflow.
- Load a data file and select spectra scans.
- Select the Select from Available Databases button, and select or add an appropriate database for the analysis.
- Select Deconvolution Settings and adjust the parameters if needed.
-
Process the data as follows:
-
Select a region on the Chromatogram to start the processing.
The resulting masses display as a list under Proteoforms.
- To change the display or specify selections for deconvolution, select Options in the Spectrum viewer.
- Adjust the Search Settings if needed.
- To zoom, select anywhere on the chromatogram and drag the mouse.
- To add a custom mass, select Add Mass.
- To search results in a database, select Search Results.
- To change the view of spectrum, select m/z, Mass, or Mass and m/z.
-
Select a region on the Chromatogram to start the processing.
-
In the Results tab, select Add to add a new set of results for the Top-Down search.
Note: Select Remove to delete a set of data you do not need for the Top-Down search. However, the last subunit cannot be removed by selecting this button.