Deconvolution Settings dialog box parameters
The following table describes the parameters you can adjust in the Deconvolution Settings dialog box.
Table 1. Deconvolution Settings dialog box parameters
Parameter
Description
Maximum Masses per Spectrum
Specifies the maximum number of deconvolution results that can be returned.
Minimum Required Charge States
Specifies the minimum number of charge states that must be found for each mass result.
Minimum Consecutive Charges
Specifies the minimum number of consecutive charge states for each mass result.
Minimum Signal-to-Noise
Specifies the minimum signal to noise ratio that a data point needs to be processed with the deconvolution.
Min/Max Mass
Specifies the mass range considered by both the charge state and the isotope deconvolution algorithms.
Min/Max Charge
Specifies the charge range considered by both the charge state and the isotope deconvolution algorithms.
Number of Peaks to Assess
Specifies the number of "top" most abundant peaks that are used to find charge state distributions.
Peak Tolerance
Specifies the allowable tolerance for other masses to be included in a charge state distribution during charge state deconvolution.
Low Number of Charge States
When enabled, reduces the minimum number of charge states required to define a mass.
Search for Additional Masses
When enabled, searches the spectra near found masses for additional related proteoform masses.
Range for Additional Masses
Specifies the search range for additional masses in m/z.