Adjust the deconvolution settings
The Interactive Deconvolution window loads with a default set of deconvolution settings. However, these might not be correct for the particular data file. The ProSight Native application is delivered with Method Templates for different types of proteins. You can apply a Method Template and then make adjustments to the specific settings.
Procedure
  1. Select Deconvolution Settings in the right pane.
    The Deconvolution Settings dialog box displays with adjustable parameters, including the charge state deconvolution, mass range, and signal to noise cutoff.
    GUID-44D5ED40-1678-438C-BEF6-EFBA842B158D-print.png
  2. Select Method Templates to open an expander that contains the list of default and saved methods.
    The currently applied Method Template is Default Auto Deconvolution. In this example, these settings are not correct for this data file, as the maximum mass in the Default Auto Deconvolution is less than the mass of the antibody in this data file.
    GUID-062A2E15-74B0-4919-96F9-98D6399C460D-print.png
  3. Select Apply next to the Default Native Antibody method template.
    The Applied checkbox is selected for the Default Native Antibody.
  4. Select OK to close the Deconvolution Settings dialog box.
  5. Select Deconvolute if the settings do not update.