Deconvolute and review data
Procedure
  1. Under the Browse button in the Intact Masses pane, select Deconvolute.
    ProSight Native performs mass deconvolution using the new settings and displays the results in the Spectrum panel.
  2. Left-click and drag across the spectrum to zoom in to all the proteoform mass peaks in one charge state, for example +26.
  3. Select the Options tab in the Spectrum panel to display additional features, such as swapping between light mode and dark mode and smoothing.
    GUID-786FB19D-2825-42AF-A1D8-5EEDA799C262-print.png
  4. Select the magnifying glass icon on the top-right of the Spectrum panel to open a side-by-side view of the m/z and deconvoluted spectrum.
    The Spectrum Viewer dialog box displays.
    GUID-AE518EB0-DAB1-4772-888B-7A0D95B6B7FA-print.png
  5. View the data grid under the Intact Masses panel to see tabular results for all of the deconvoluted proteoforms detected, including mass, intensity, and delta mass.
    GUID-C012CA22-D4BE-4C46-A384-4B307D09D3BB-print.png