The following table describes the commands available in the Theoretical Protein/Peptide Manager.
Command | Description |
---|---|
Import Protein Fasta | Opens a dialog box where you can browse and select a FASTA file to add a protein sequence to the Theoretical Protein/Peptide Manager. |
Export | Exports all or selected results in the Theoretical Protein/Peptide Manager Results table as an Excel ( |
Save Results As | Saves all theoretical component results in the Theoretical Protein/Peptide Manager Results table as an |
Save As a Peptide Workbook | Saves all or selected component results in the Theoretical Protein/Peptide Manager Results table to a workbook file ( |
Load Results | Opens a dialog box where you can browse and select |