Right-clicking anywhere on the HDX protection factor plot on the Hydrogen Deuterium Exchange page opens a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Reset Scale | Restores the original scale to the HDX protection factor plot. |
Copy | Copies the image viewable in the HDX protection factor plot. |
Add to Report | (Ardia Platform mode only) Sources the HDX protection factor plot for the report. |