The following table describes the types of information in the Results table on the Host Cell Protein Quantitation Results page.
Parameter | Description |
|---|---|
Field Chooser | Displays the Field Chooser dialog box so that you can select the columns to display in the lower section of the Modification Results pane. |
(Check box) | Select or clear the checkbox in the table header row to select/deselect all rows in the lower section. You can also select the checkbox in individual rows for export. |
Protein ID | Displays the number assigned to a specific HCP in the analysis. If you sort/filter results in the table, the assigned component number will remain unchanged for a component, although its row number within the table may change. |
Protein Description | Displays the name and ID information of the HCP. |
Protein Mass | Displays the mass of the HCP. |
Total # of Peptides | Displays the number of peptides associated with the HCP. |
% Confidence | Displays the confidence score for the HCP assignment as a percent. |
Score | Displays the confidence score for the HCP assignment. |
Average MS area for raw_data_file_name | Displays the average MS area for the HCP for a particular condition/raw data file. |
%CV Peptides for raw_data_file_name | Displays the coefficient of variation (CV) for the HCP for a particular condition/raw data file. %CV = (S ÷ A) × 100, where
|
Peptides for raw_data_file_name | Displays the peptide sequences associated with the HCP for a raw data file in the experiment. The number of peptide sequences listed in this column equals the value listed in the Total # of Peptides column. |