Right-clicking the Results table on the Host Cell Protein Quantitation page opens a shortcut menu with the commands listed in the following table.
Host Cell Protein Quantitation Results table shortcut menu
Command
Description
Export All
Exports data for all results in the Host Cell Protein Quantitation Results table to an Excel workbook (.xlsx).
Export Checked
Exports data for only the selected results in the Host Cell Protein Quantitation Results table to an Excel workbook (.xlsx).
Add to Report
(Ardia Platform mode only) Sources all or selected results for the report.