Right-click the Results table on the Coverage page to open a shortcut menu with the commands listed in the following table.
Table Results table shortcut menu
Command
Description
Show Protein Coverage Map Parameters
Opens the Coverage Map Options dialog box to modify the peptide recovery parameters and the number of residues per row displayed on the Protein Sequence Coverage Map.
See Modify Sequence Coverage Map parameters.
Export
Exports all data (at the protein- and raw data file-levels) in the Coverage Results table to an Excel workbook (.xlsx).