The following table summarizes the types of fragment ions that appear in the spectra in the MS2 Spectra pane.
Fragment | Description | Color |
Ion series | ||
a | A ion with a charge on the N-terminal side | Light blue |
b | B ion with a charge on the N-terminal side | Dark blue |
c | C ion with a charge on the N-terminal side | Dark green |
c· | C ion with a charge on the N-terminal side, one less proton (c−1.0078) | Light green |
x | X ion with a charge on the C-terminal side | Orange |
y | Y ion with a charge on the C-terminal side | Red |
z | Z ion with a charge on the C-terminal side | Purple |
z· | Z ion with a charge on the C-terminal side, one more proton (z+1.0078) | Light purple |
z·· | Z ion with a charge on the C-terminal side, two more protons (z+2(1.0078)) | Lighter purple |
Neutral losses | ||
−H2O | Fragment that has lost water (−18 Da) | |
−NH3 | Fragment that has lost ammonia (−17 Da) | |
Post-translational modifications (PTMs) | ||
H3PO4 | Phosphorylation loss (designated −P) | |
Other | ||
Immonium | Immonium ion | |
M | Precursor ion, M Neutral loss, for example:
| |
R | Neutral loss from arginine with a minus mass value, for example, R-44 | |
Glycans | There are several types of glycan labels:
Each type can have a charge associated with it. Glycan labels include capital letters to distinguish them from ion series labels. See Appendix: Glycans. |