The following table describes the parameters in the Charge State Results table on the Process and Review page for Peptide Mapping Analysis experiments.
Table Charge State Results table for Peptide Mapping Analysis experiments
Column
Description
Columns marked with an asterisk (*) are only present in the Results table when the certain features are enabled in the experiment, as detailed below.
+/– (button)
Shows/hides raw data file-specific (lower-level) information related to the currently selected component row.
Row number (bolded)
Displays the row number of the table.
This number corresponds to the number of currently visible rows in the table; it is not assigned to a specific component. If you sort/filter results in the table, the row numbers will remain unchanged while the filtered/sorted components that populate the rows change
(Check box)
Selects/deselects a component row.
To select/deselect the checkboxes for all component rows at once, select or clear the checkbox in the column header.
If you filter the table, the following occurs:
  • In a filtered view, selecting all checkboxes affects only the filtered (currently visible) component rows.
  • In a filtered view, deselecting all checkboxes affects all the component rows in the table, currently visible or not.
Level
Indicates the level of information displayed as either component level (top-level) or raw data file-specific (lower-level).
No.
Displays the number assigned to a specific component in the analysis.
Component number is not related to table row number (see above).
If you sort/filter results in the table, the assigned component number will remain unchanged for a component, although its row number within the table may change.
*Alias Name
Identifies a component using the custom chain root aliases applied to theoretical digestion results. Only components that match a theoretical digestion component have an alias name listed in this column.
You can edit the entries in this column by placing your cursor in the Alias Name box for the currently selected component row and typing a new alias name. If you added labels to your chromatogram, you can update the labels to reflect the changes you made in the Alias Name column by selecting the “Refresh Alias” button at the top of the Chromatogram pane.
*This column is only visible when the Customize Results checkbox is selected on the Customize Results subpage of the Parameters page.
Identification
Identifies key characteristics of an identified peptide component and any applied modifications.
For example, 7:T95-R119 = 2544.30062m(C98+Carbamidomethylation) indicates the following:
  • The protein chain ID number (7)
  • The first amino acid in the peptide sequence and its position (T95)
  • The last amino acid in the peptide sequence and its position (R119)
  • The mass of the neutral, unmodified peptide (2544.30062m)
  • Any modifications and their respective site locations (C98+Carbamidomethylation)
If a component is not identified, this column is empty (blank).
If a component contains a disulfide bond, this column displays both peptide sequences involved in the bond, separated by a forward slash (“/”). If the disulfide bond involves peptide sequences from different protein chains, the respective parent protein chain ID numbers are listed in parentheses. For example, the peptide sequence WWCNDGR(2)/GCRL(3) indicates that there is a disulfide bond between the WWCNDGR residue on protein chain 2 and the GCRL residue on protein chain 3.
In a targeted Peptide Mapping Analysis, the Comment column of the table may suggest a possible identification for a component, even if the Identification column is empty.
In some situations, a combination of low resolution, large component mass, or high component charge state may prevent the application from resolving the monoisotopic mass (m) of a component. In this situation, the average mass (a) will be used in the Identification column notation.
For example, 1128.27a is the average mass for a given component. Due to low resolution, the application cannot calculate the monoisotopic mass (m) for this component and displays a value of 0 in the Mono Mass Exp. column of the table. Therefore, in the Identification column, the average mass 1128.27a is used in the notation, rather than the monoisotopic mass (m).
Peptide Sequence
Displays the peptide sequence constituting an identified component.
If a component is not identified, this column is empty (blank).
If a component contains a disulfide bond, this column displays both peptide sequences involved in the bond, separated by a forward slash (“/”). If the disulfide bond involves peptide sequences from different protein chains, the respective parent protein chain ID numbers are listed in parentheses.
For example, the peptide sequence WWCNDGR(2)/GCRL(3) indicates that there is a disulfide bond between the WWCNDGR residue on protein chain 2 and the GCRL residue on protein chain 3.
*Start #
Displays the position of the first amino acid residue in a theoretical digestion component.
*This column is only visible when the Customize Results checkbox is selected on the Customize Results subpage of the Parameters page.
*End #
Displays the position of the last amino acid residue in a theoretical digestion component.
*This column is only visible when the Customize Results checkbox is selected on the Customize Results subpage of the Parameters page.
Mod
Displays the type(s) of modification(s) associated with an identified component.
For a description of the notation used in the "Mod" column, see Modification notation.
If there is no modification, this column displays “None”.
If a component is not identified or if the Seq. Variant column displays an amino acid substitution, this column is empty (blank).
If a component is identified but does not adhere to the expected activity of the protease you selected, this column displays “Nonspecific”.
The modifications listed in this column may include the variable modifications assigned during processing, nonspecific modifications, and other modifications determined automatically by the application. In this case, the application lists variable modification first, followed by static modifications in parentheses. Multiple modifications are separated by commas (“,”).
Site
Displays the location of the modification(s) listed in the Identification/Mod columns or the amino acid substitution(s) listed in the “Seq. Variant” column for an identified component.
For example, if the Identification column displays 7:T95-R119 = 2544.30062m(C98+Carbamidomethylation) for a component, the site of the carbamidomethylation modification is C98.
If a component is not identified, this column in empty (blank).
The application lists the sites for variable modifications first, followed by the sites for static modifications in parentheses. Multiple modifications are separated by commas (“,”).
If the application cannot identify the exact site of a modification or if there are multiple candidate residues within a component, the application indicates an approximate modification site with a tilde (̴̴̴~).
If a component involves a disulfide bond, this column displays a forward slash (/) to separate the sequences involved in the bond.
For example, if the Identification column displays 1:C6-R14/N44-R68 = 3662.03A[1ss] and within the N44-R68 sequence, there is cysteine residue at position 64, the site for this disulfide bond is 1:C6/1:C64.
*Seq. Variant
Displays any sequence variants (amino acid substitutions) associated with an identified component.
If a component is not identified or does not have any sequence variants, this column is empty (blank).
*This column is only visible when the Search for Amino Acid Substitutions parameter on the Identification subpage of the Parameters page is set to “Single Base Change” or “All Substitutions”.
∆ ppm
Displays the mass difference between the theoretical monoisotopic mass (Theor. Mass) of the component and the experimental monoisotopic mass (Mono Mass Exp.) of an identified component.
∆ ppm = [(Mono Mass Exp. – Theor. Mass) ÷ Theor. Mass] × 106
If a component is not identified, this column is empty (blank).
If the application cannot confidently determine the Mono Mass Exp. value for a component, it will use the Avg Mass Exp. to calculate ∆ ppm.
Conf. Score
Indicates how closely the predicted and experimental MS spectra match for an identified component using a percentage scale (between 0 and 100%).
  • A confidence score of 0% indicates a poor match between the predicted and experimental spectra.
  • A confidence score of 100% indicates a very good (although not necessarily exact) match between the predicted and experimental spectra.
If a component is not identified, this column is empty (blank).
*Comet E-value
Displays the E-value (expectation score) for a component identified using the host cell protein database (Comet) search feature.
E-value is used in the calculation of a component’s PEP (posterior error probability) score.
This value is displayed in scientific notation to two decimal places.
*This column is only visible when you enable the host cell protein database searching feature on the Host Cell Protein subpage of the Parameters page.
Best ASR
Displays the best average structural resolution (ASR) value available for an identified component, considering all of the individual raw data files used in the experiment. The value displayed in this column is the best ASR value (value closest to 1.0) out of all the raw data files for the component.
ASR = (Number of Amino Acids)/(Number of Bonds Found + 1)
Each raw data file used in the experiment has its own associated ASR which can be viewed by expanding the selected component row to view raw data file-specific information.
The Best ASR value is also displayed on the Fragment Coverage Map of the Mapping page.
ID Type
Indicates the type of MS data the application used to identify a component.
  • Component identified using MS/MS scan data.
  • Full: Component identified using full-scan data.
  • MS2/Full: Component identified using both MS/MS and full-scan data.
  • Associated: Component identity inferred from other component identities consistent across multiple raw data files.
  • Note: "Associated" components do not have a corresponding MS2 spectrum.
If a component is not identified, this column is empty (blank).
MS/MS data provides more confidence than full-scan data in identifying a component.
When the experiment involves a single raw data file, this column displays the same ID Type at both the component level and raw data file-specific level.
When the experiment involves multiple raw data files, the ID Type used for each individual file can be viewed by expanding the selected component row to view raw data file-specific information.
Integration Type
Indicates the type of integration used to calculate the MS area for a component.
  • “Automatic” indicates that the application automatically determined the integration.
  • “Manual” indicates that the user adjusted retention times and created the integration.
RT
Displays the peak retention time (RT) for a component.
If the experiment involves multiple raw data files, the RT displayed in this column is the retention time associated with the first raw data file.
Additionally, at the raw data file-specific information level, RT Start and RT End columns display the start and end time points of the retention time range for each individual raw data file.
M/Z
Displays the mass-to-charge ratio of the theoretical monoisotopic mass of a component.
Charge St.
Displays the charge state of a component.
Charge state refers to the imbalance between the number of protons and the number of electrons in a molecular species (or an ion adduct).
Mono Mass Exp.
Displays the experimental monoisotopic mass for a component.
This mass is based on the exact mass of the most abundant isotope of each element in a molecule.
If the application cannot confidently determine the Mono Mass Exp. of a component, this column displays a value of “0”.
Avg. Mass Exp.
Displays the experimental average mass for a component.
This mass is based on the average atomic weight of each element in a molecule.
Theor. Mass
Displays the theoretical monoisotopic mass for an identified component.
If a component is not identified, this column is empty (blank).
Missed Cleavages
Displays the number of missed cleavages involved in producing an identified component; intrinsically related to the activity of the protease selected for the experiment.
*Host Cell Protein
Indicates if a component was identified using the using the host cell database search (Comet).
  • “No” indicates that component was identified by the application.
  • “Yes” indicates that the component was identified using the host cell database search (Comet).
*This column is only visible when you enable the host cell protein database searching feature on the Host Cell Protein subpage of the Parameters page.
Protein
Displays the default protein name (derived from the FASTA file) associated with an identified component.
If you use de novo sequencing to identify a component, this column displays “De Novo”.
If a component is not identified, this column is empty (blank).
*Ratio (Condition/Reference Condition)
Displays the following ratio for a component:
Ratio = Avg MS Area (Condition)/Avg MS Area (Reference Condition)
The Results table includes a Ratio column for each condition (excluding the reference condition) specified in the experiment.
If the reference condition value is 0 for a component, this column displays “999.99”.
*This column is only visible when multiple raw data files and conditions are used in an experiment.
*Max Cond.
Displays the experiment condition with the highest ratio value.
See the Ratio (Condition/Reference Condition) column for more information.
*This column is only visible when multiple raw data files and conditions are used in an experiment.
*Min. Cond.
Displays the experiment condition with the lowest ratio value.
See the Ratio (Condition/Reference Condition) column for more information.
*This column is only visible when multiple raw data files and conditions are used in an experiment.
*Max MS Area
Displays the highest MS area determined for a component, considering all of the conditions specified in an experiment.
*This column is only visible when multiple raw data files and conditions are used in an experiment.
MS Area or Avg. MS Area: Condition
For an experiment with a single raw data file, displays the MS area for a component for a particular condition.
For an experiment with multiple raw data files, displays the average MS area for a component for a particular condition, considering all of the individual raw data files used in an experiment.
*%CV: Condition
Displays the coefficient of variation (CV) for a component for a particular condition.
%CV = (S ÷ A) × 100, where
  • S is the standard deviation of the MS Area values from all of the individual raw data files that belong to a particular experiment condition.
  • A is the average of those MS Area values
The Results table includes a %CV column for each condition specified in the experiment.
*This column is only visible when multiple raw data files and conditions are used in an experiment.
Comment
Editable. You can enter comments (up to 128 alphanumeric characters and/or symbols) related to the selected component in this column.
Comments entered in the Results table on the Process and Review page automatically appear in the Comment column of the Components table on the Modification Summary page (and vice versa).
In a targeted Peptide Mapping Analysis, the Comment column of the table may suggest a possible identification for a component, even if the Identification column is empty.
*Protein Family
Displays the total number of proteins that contain the selected peptide sequence found when searching the host cell protein database (Comet).
*This column is only visible when you enable the host cell protein database searching feature on the Host Cell Protein subpage of the Parameters page.
*Other Protein Description
Displays a list of all of the proteins that contain the selected peptide sequence found when searching a host cell protein (HCP) database (Comet).
Proteins are listed by their accession numbers (with descriptions); multiple proteins are separated by a comma (“,”).
*This column is only visible when you enable the HCP search feature on the Host Cell Protein subpage of the Parameters page.