Display a single chromatogram plot type for multiple raw data files - BioPharma Finder Software - BioPharma Finder 5.4 Peptide Mapping Analysis User Guide - View the chromatograms - View Peptide Mapping Analysis results - Display a single chromatogram plot type for multiple raw data files - Chromatogram plot types - Process and Review page - BioPharma Finder Software - BioPharma Finder Software
Ardia Platform > Software > BioPharma Finder Software
Desktop Software > BioPharma Finder Software
Document Type
User Guide
Software version
5.4
Prerequisites
You have loaded the experiment results are viewing the Process and Review page.
Procedure
On the Process and Review page, right-click the Chromatogram pane and select Select Chromatogram.
The Select Chromatogram dialog box opens.
Your raw data file selections in the Select Chromatogram dialog box control the chromatogram plots. To return the control of the chromatogram plots to your selections in the Results table, you must clear all raw data file selections in this dialog box and then select OK. Otherwise, proceed.
If you select only one plot type from the list on the left, you can select multiple raw data files from the list on the right.
Conversely, to select multiple plot types, you must select only one raw data file.
On the left side of the dialog box, select the single chromatogram plot type you want to view.
On the right side of the dialog box, select the raw data files whose chromatograms you want to display from the list.
To select all raw data files in the list, select the checkbox in the column header.
Figure Selecting a chromatogram plot type for multiple raw data files
Select OK to update the chromatogram plots.
The Chromatogram pane displays the specified chromatograms for all of the selected raw data files.
Figure Base peak chromatograms (BPC) displayed for many different raw data files