You can change the Sequence Coverage Map parameters to modify the peptide recovery parameters and the number of residues displayed per row.
Prerequisites
- You have loaded the experiment results are viewing the Coverage page under the Mapping tab.
Procedure
- On the Coverage page, right-click anywhere in the Coverage Results table and select Show Protein Coverage Map Parameters from the shortcut menu.
- The Coverage Map Options dialog box opens.
Figure Coverage Map Options dialog box NOTE
If you define specific components for the Sequence Coverage Map, you can update only the "Residues per Row" parameter in the Coverage Map Options dialog box.
The other parameters are read-only.
- Enter the following settings:
- Maximum Peptide Mass: Type a value to specify the maximum peptide mass to be included in the Sequence Coverage Map.
- Minimum Confidence: Type a value between 0.00 and 1.00 to set the minimum confidence level for a peptide assignment on a 0 to 100% scale, with 1.00 being the highest confidence. The application automatically converts this value to a percentage value between 0.00% and 100.00%.
- If you set the Search by Full MS Only option to "Yes" in the processing method, set this value to 0.00%. Otherwise, you will not see any coverage.
- Residues per Row: Type the maximum number of amino acids (residues) to display in each row of the Sequence Coverage Map.
- Minimum Recovery: Type a value between 0.00 and 1.00 to specify the minimum recovery threshold on a 0 to 100% scale, with 1.00 being the highest recovery. The application automatically converts this value to a percentage value between 0.00% and 100.00%.
- A peptide must have a recovery value higher than this threshold to be included in the coverage map for a peptide assignment.
- Minimum Relative Recovery of Overlapping Peptides: Type a value between 0.00 and 1.00 to specify the minimum relative abundance threshold on a 0 to 100% scale, with 1.00 being the highest threshold. The application automatically converts this value to a percentage value between 0.00% and 100.00%.
- A peptide must have a relative abundance value higher than this threshold to be included in the Sequence Coverage Map.
- Click OK.
- The Sequence Coverage Map updates based on your settings.
IMPORTANT
The parameter settings in the Coverage Map Options dialog box are not related to the chromatogram shading settings specified for the Coverage chromatogram.
For more information on the shading settings used in the Coverage chromatogram, see Modify the shading parameters.