You can specify which components to include in the Sequence Coverage Map on the Coverage page by selecting specific components in the Results table on the Process and Review page.
Prerequisites
- You have loaded the experiment results are viewing the Coverage page under the Mapping tab.
Procedure
- From the Coverage page, click the Process and Review tab in the navigation bar.
- In the Results table, select or clear the check boxes for the components that you want to include in or exclude from the Sequence Coverage Map on the Coverage page.
- Navigate back to the Sequence Coverage Map on the Coverage page by clicking the Mapping > Coverage tab in the navigation bar.
- The Sequence Coverage Map now shows only the components selected on the Process and Review page.
- The map title now displays "Sequence Coverage MapāUser Defined".
- To return to the default Sequence Coverage Map, repeat the process above, but in the Results table on the Process and Review page, select the check box in the table header row to clear all of the check boxes and deselect all components.