The BioPharma Finder application uses the kinetic model to predict peptide MS/MS (MS2) spectra. In the MS2 Spectra pane on the Process and Review page, you can change the parameters to regenerate the predicted spectrum using the model.
Prerequisites
- For a Peptide Mapping Analysis experiment, you have processed your experiment and are viewing the results on the Process and Review page.
Procedure
- On the Process and Review page, select the MS2 Spectra subtab.
- Select a row in the Results table that displays "MS2" in the ID Type column.
- Right-click the MS2 Spectra pane and select Predict Peptide MS/MS (Kinetic Model).
- The application opens the Predict Peptide MS/MS (Kinetic Model) dialog box.
Figure Predict Peptide MS/MS (Kinetic Model) dialog box - In the Sequence box, type the one-letter codes for a peptide sequence.
- See Amino acid letter codes.
- Use uppercase when typing the one-letter codes.
- In addition to the standard one-letter peptide codes, the following special peptide codes are accepted:
- •D for deamidated asparagine
- •J for carboxymethylated cysteine
- •U for carboxyamidomethylated cysteine
- •O for oxidized methionine
- •S for phosphorylated serine
- Additionally, you can indicate modifications by typing the modification in parentheses after the sequence.
- For example, AADECFGHK(C5+250)(H8−9) means the cysteine at position 5 is modified by +250 u and the histidine at position 8 is modified by −9 u.
- Another example: AANASAA(N3+A2G0F) means the asparagine at position 3 is glycosylated with glycan A2G0F.
- To define a disulfide bond, type the bond in parentheses.
- For example: ADCAGHTYCHPEK(C3−C9) means the cysteine at position 3 and the cysteine at position 9 are linked by a disulfide bond.
- Enter values for the following parameters:
- To set the charge state, the isolation width used for fragmentation, the resolution at m/z = 400, and the activation time (ms), enter a value in their boxes.
- To define a fragmentation method, select from the available options in the list.
- To select an instrument, select an instrument name from the list.
- To define the collision energy or reagent target, enter a value in the box.
- Specify normalized collision energy as a percentage value (%) or specify the reagent ion target value for the ETD fragmentation method.
- Select OK to view the predicted MS/MS spectrum.
- The application displays the changes in the predicted spectrum in the MS2 Spectra pane.
- See View the predicted and experimental MS/MS spectra.
NOTE
For components identified as dimers, the resulting predicted MS/MS spectrum using the kinetic model will not be correct.
Figure Predicted MS/MS spectrum after applying the kinetic model