In the Peptide Mapping Analysis workflow you can run both non-targeted and targeted experiments:
In a non-targeted experiment, the BioPharma Finder application identifies components using the framework of a designated protein sequence, if specified.
Figure Non-targeted Peptide Mapping Analysis experiment workflow
Non-targeted Peptide Mapping Analysis experiment workflow
In a targeted experiment, the application identifies components using the framework of a target peptide workbook, which contains component results saved from an initial non-targeted experiment.
Figure Targeted Peptide Mapping Analysis experiment workflow
Targeted Peptide Mapping Analysis experiment workflow