Right-clicking the Components table on the Modification Summary page opens a shortcut menu with the commands listed in the following table.
Components table shortcut menu
Command
Description
Export All Components
Exports all data in the Components table to an Excel workbook (.xlsx).
Export Checked Components
Exports data only for the selected components in the Components table to an Excel workbook (.xlsx).
Save As Peptide Workbook
Saves all or selected component results to a workbook (.wbpf) that is Chromeleon-compatible and used for targeted experiments.
Add to Report
(Ardia Platform mode only) Sources all or selected results for the report.