Right-clicking the Modification Results table on the Modification Summary page opens a shortcut menu with the commands listed in the following table.
Modification Results table shortcut menu
Command
Description
Set Summary Options
Opens the Summary Options dialog box so that you can set new values for the Peptide Intensity (%), Charge State Minimum Intensity (%), and Minimum Modification Level (%) columns in the upper section of the Modification Results table.
Export All Modifications to Excel
Exports data for all results in the Modification Results table to an Excel workbook (.xlsx).
Export Checked Modifications to Excel
Exports data for only the selected results in the Modification Results table to an Excel workbook (.xlsx).
Add to Report
(Ardia Platform mode only) Sources all or selected results for the report.