You can export the data in the Modification Results table as an Excel worksheet (.xlsx).
Prerequisites
  1. You have loaded the experiment results are viewing the Modification Summary page under the Mapping tab.
Procedure
  1. On the Modification Summary page, right-click anywhere in the Modification Results table and select one of the following options on the shortcut menu:
  2. Export All Modifications to Excel to export all of the results in the table as an Excel worksheet (.xlsx).
  3. Export Checked Modifications to Excel to export only the selected results as an Excel worksheet (.xlsx).
  4. The Save As dialog box opens.
  5. To select specific results, select the checkbox for a modification row in the lower section of the Modification Results table.
  6. To select or deselect all of the rows, select or clear the checkbox in the table header row.
  7. Browse to the file location where you want to save your results and type a file name.
  8. Select Save.
  9. The application saves the results as an Excel worksheet (.xlsx).