On the Target Peptide Workbook page, you can import, export, and delete workbooks using the Workbook Manager.
TIP
You can access the Workbook Manager by selecting the Target Peptide Workbook tab on the navigation bar within the Peptide Mapping Analysis workflow.
The following table describes how to perform common Workbook Manager tasks.
Task | Actions | Result |
---|---|---|
Edit a workbook | In the Workbook Manager table, select the checkbox for a workbook row and then select Edit on the command bar. | The Workbook Editor page opens. |
Import a workbook | Select Import on the command bar. Then, in the Open dialog box, locate and select the workbook files (. | A confirmation message appears. Select OK to continue. The imported workbooks are added to the Workbook Manager table. If an imported workbook has the same name as an existing workbook in the table, the application appends " |
Export a workbook | In the Workbook Manager table, select the checkbox for one or more workbook rows and then select Export on the command bar. Then, in the Save As dialog box, browse to the folder location where you want to save the workbook files ( | A confirmation message appears. Select OK to continue. The selected workbooks are saved in the specified file location. |
Delete a workbook | On the Workbook Manager table, select one or more workbook rows (not checkboxes) and then either select Delete on the command bar or press the DELETE key. |
A confirmation message appears. Select OK to continue. The selected workbooks are removed from the Workbook Manager table and permanently deleted from the application. |