The following table lists the O-linked glycans that the application supports for the O-glycan-specific search in peptide mapping analysis.
Table O-Linked glycans
#
Glycan
Monoisotopic mass
1
GalNAc
203.079
2
GalNAc-3G
365.132
3
GalNAc-3GnG
568.212
4
GalNAc-3SG
656.228
5
GalNAc-6GGn
568.212
6
GalNAc-6GGn-3G
730.264
7
GalNAc-6GGn-3GnG
933.344
8
GalNAc-6GGn-3SG
1021.36
9
GalNAc-6Gn
406.159
10
GalNAc-6Gn-3G
568.212
11
GalNAc-6Gn-3GnG
771.291
12
GalNAc-6Gn-3SG
859.307
13
GalNAc-6S
494.175
14
GalNAc-6S-3G
656.228
15
GalNAc-6S-3GnG
859.307
16
GalNAc-6S-3SG
947.323