The following table lists the N-linked glycans, sorted by human host cell-line type, that are included in the N-glycan-specific search.
In the table header, "Mono mass" refers to monoisotopic mass, "Man" refers to mannose, "Fuc" refers to fucose, and "Bis Gn" refers to bisecting glycoprotein.
Table N-Linked glycans with a human host cell-line type
#
Glycan name
Antenna
Formula
Mono mass
Man
Fuc
Bis Gn
1
2
3
4
1
M3
C34H56N2O25
892.317
3
0
0
2
M4
C40H66N2O30
1054.37
4
0
0
3
M5
C46H76N2O35
1216.42
5
0
0
4
M6
C52H86N2O40
1378.48
6
0
0
5
M7
C58H96N2O45
1540.53
7
0
0
6
M8
C64H106N2O50
1702.58
8
0
0
7
M9
C70H116N2O55
1864.63
9
0
0
8
A1G0
Gn-
C42H69N3O30
1095.4
3
0
0
9
A1G0F
Gn-
C48H79N3O34
1241.45
3
1
0
10
A2G0
Gn-
Gn-
C50H82N4O35
1298.48
3
0
0
11
A2G0B
Gn-
Gn-
C58H95N5O40
1501.56
3
0
1
12
A2G0F
Gn-
Gn-
C56H92N4O39
1444.53
3
1
0
13
A2G0FB
Gn-
Gn-
C64H105N5O44
1647.61
3
1
1
14
A1G1
G-Gn-
C48H79N3O35
1257.45
3
0
0
15
A1G1F
G-Gn-
C54H89N3O39
1403.51
3
1
0
16
A2G1
G-Gn-
Gn-
C56H92N4O40
1460.53
3
0
0
17
A2G1B
G-Gn-
Gn-
C64H105N5O45
1663.61
3
0
1
18
A2G1F
G-Gn-
Gn-
C62H102N4O44
1606.59
3
1
0
19
A2G1FB
G-Gn-
Gn-
C70H115N5O49
1809.67
3
1
1
20
A2G2
G-Gn-
G-Gn-
C62H102N4O45
1622.58
3
0
0
21
A2G2B
G-Gn-
G-Gn-
C70H115N5O50
1825.66
3
0
1
22
A2G2F
G-Gn-
G-Gn-
C68H112N4O49
1768.64
3
1
0
23
A2G2FB
G-Gn-
G-Gn-
C76H125N5O54
1971.72
3
1
1
24
A1S1
S-G-Gn-
C59H96N4O43
1548.54
3
0
0
25
A1S1F
S-G-Gn-
C65H106N4O47
1694.6
3
1
0
26
A2S1G0
S-G-Gn-
Gn-
C67H109N5O48
1751.62
3
0
0
27
A2S1G0B
S-G-Gn-
Gn-
C75H122N6O53
1954.7
3
0
1
28
A2S1G0F
S-G-Gn-
Gn-
C73H119N5O52
1897.68
3
1
0
29
A2S1G0FB
S-G-Gn-
Gn-
C81H132N6O57
2100.76
3
1
1
30
A2S1G1
S-G-Gn-
G-Gn-
C73H119N5O53
1913.68
3
0
0
31
A2S1G1B
S-G-Gn-
G-Gn-
C81H132N6O58
2116.76
3
0
1
32
A2S1G1F
S-G-Gn-
G-Gn-
C79H129N5O57
2059.73
3
1
0
33
A2S1G1FB
S-G-Gn-
G-Gn-
C87H142N6O62
2262.81
3
1
1
34
A2S2
S-G-Gn-
S-G-Gn-
C84H136N6O61
2204.77
3
0
0
35
A2S2B
S-G-Gn-
S-G-Gn-
C92H149N7O66
2407.85
3
0
1
36
A2S2F
S-G-Gn-
S-G-Gn-
C90H146N6O65
2350.83
3
1
0
37
A2S2FB
S-G-Gn-
S-G-Gn-
C98H159N7O70
2553.91
3
1
1
38
Gn
C8H13N1O5
203.079
0
0
0
39
GnF
C14H23N1O9
349.137
0
1
0