You can view the host cell protein (HCP) search results for your experiment on the Host Cell Quantitation page. The Host Cell Quantitation page quantifies the HCP contaminants detected in your experiment. This page is active only when you enable the HCP search feature on the Host Cell Protein page of the processing method editing wizard.
Procedure
- Open the results of your experiment from the Queue page or from the Load Results page.
- See Open results.
- The Process and Review page opens by default.
- In the navigation bar, select the Mapping tab.
- By default, the application opens the Coverage page under the Mapping tab.
- In addition to the Coverage page, the Mapping tab also contains the Modification Summary, Hydrogen Deuterium Exchange (for an HDX experiment), and Host Cell Protein Quantitation (if the HCP search feature is enabled) pages.
- On the Mapping page, select the Host Cell Protein Quantitation subtab.
- The Host Cell Protein Quantitation page opens, displaying the Host Cell Protein Quantitation Results and Components tables on the left side and the Host Cell Protein Relative Quantitation plot on the right.
Figure Host Cell Protein Quantitation page