In addition to the Process and Review page, you can view the results of your Peptide Mapping Analysis experiment on the Coverage page, under the Mapping tab. The Coverage page provides detailed information regarding the coverage of the identified and unidentified proteins in the experiment.
Procedure
- Open the results of your experiment from the Queue page or from the Load Results page.
- See Open results.
- The Process and Review page opens by default.
- In the navigation bar, click the Mapping tab.
- By default, the application opens the Coverage page under the Mapping tab, which displays the color-coded chromatogram in the Chromatogram pane, the Sequence Coverage Map in the Protein Coverage Map pane, and the Results table, all denoting the identified and unidentified proteins in the experiment.
- In addition to the Coverage page, the Mapping tab also contains the Modification Summary, Hydrogen Deuterium Exchange (for an HDX experiment) and Host Cell Protein Quantitation (if the HCP search feature is enabled) pages.
Figure Coverage page