The Peptide Match ID pane on the Process and Review page provides detailed fragment information for a selected component when MS/MS data is available.
Procedure
- Open the results of your experiment from the Queue page or from the Load Results page.
- See Open results.
- The Process and Review page opens by default and displays the results of your Peptide Mapping Analysis experiment, including the peptide match ID results.
- On the top left side of the Process and Review page, select the Peptide Match ID Details tab (tabbed together with the Chromatogram pane).
Figure Peptide Match ID pane - The Peptide Match ID Details pane opens and contains two expandable sections: Select Fragments and Component Summary.
- The Select Fragments section contains the following parameters: Ion Series, Neutral Losses, and Internal Fragments.
- Select a component row in the Results table to edit the Select Fragments and Component Summary sections of the Peptide Match ID Details tab.
- In the Select Fragments section, select the checkbox for the fragments you want to display in the Component Summary table below and select the Apply button.
- The Component Summary table updates according to the parameters specified in the Select Fragments section.
- On the top right side of the page, select the MS2 Spectra tab.
Figure MS/MS spectrum containing fragment information for a selected component - The MS/MS spectrum appears for the component selected in the Results table, with the selected fragment types labeled.
- At the top of the MS2 Spectra pane, the Activation list provides the available fragmentation types and the Resolution list provides the available resolution types for the MS/MS spectrum.
- Edit the Select Fragments section as necessary to view the Component Summary results and fragment-labeled MS/MS spectra for the selected component.