Right-click the Results table on the Process and Review page to open a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Filters | Manages the filters in the Results table. |
Apply | Applies all of the filters from a saved file to the table. |
Save As | Saves all of the filters to a file with the .cfg extension. |
Clear All | Clears all filters from the table. |
Export All Components | Exports all data in the Results table to an Excel workbook (.xlsx), text file (.csv), Mascot generic format (.mgf), or BioPharma Finder (.bpf) file. |
Export Checked Components | Exports data only for the selected components in the Results table to an Excel workbook (.xlsx), text file (.csv), or BioPharma Finder (.bpf) file. |
Create .mgf File | Creates an .mgf file that the Mascot search engine can read. |
Run De Novo Processing/Cancel De Novo Processing | Starts or cancels the de novo sequencing for a particular component. |
Show Component Information | Displays information for an identified component, including other possible identifications from de novo sequencing. |
Save As Peptide Workbook | Saves all or selected component results to a workbook (.wbpf) that is Chromeleon-compatible and used for targeted experiments. |
Add to Report | (Ardia Platform mode only) Sources all or selected component results for the report. |