For a Peptide Mapping Analysis with multiple raw data files, you must enter the experimental conditions for the files, including a reference condition, on the Peptide Mapping Analysis page before processing the experiment.
After processing, you can change this reference condition by selecting a different condition from the drop-down list at the far right of the Results table.
The application automatically recalculates and updates the Ratio (Condition/Reference Condition), Max Condition, and Min Condition columns in the Results table on the Process and Review page and the Components table on the Modification Summary page.