When the BioPharma Finder application completes the processing of your experiment, you can open the results of your analysis from the Queue page or from the Load Results page.
The Process and Review page opens by default, displaying the Results tables, chromatograms, trend ratio and trend MS area plots, hydrogen deuterium exchange (HDX) plots, peptide fragment coverage and protein sequence maps, and full-scan MS and MS/MS spectra, including predicted kinetic MS/MS spectra.
Figure Process and Review page for Peptide Mapping Analysis
Process and Review page for Peptide Mapping Analysis