The table below lists all known issues that exist in the Orbitrap Hybrid Series 5.1 Instrument Control Software release.
Table Known Issues
ID
Severity
Title
153093
3 - Medium
Divert Valves need to be Configured for USB Control.
Remedy: The divert valves can be controlled through interfacing with a computer. Download the MX Series II™ Control Software Program to configure the divert valves for Orbitrap Exploris: www.idex-hs.com/literature-tools/software-drivers/
153505
3 - Medium
Installer blocks workstation installation (Method Editor standalone) if there is already a Full Installation on the data system PC.
Remedy: Run Workstation installation on a different computer. Installer supports simultaneous Workstation installation of all Orbitrap Exploris instrument models.
153665
3 - Medium
Orbitrap Exploris 480 DVD ISO image: Software Uninstallation Error.
154441
3 - Medium
Tune: Display imperfect on 4 k monitor.
153627
3 - Medium
Tune: Under certain conditions, wrong number of errors and warnings is shown.
828949
3 - Medium
Tune: Graph for Mass Calibration in Tune gets replaced by "Adjust HCD cell pressure to standard pressure" when running from Intact Protein Mode with Standard pressure setting
153432
2 - High
UI: Method Editor: MIPS filter incompatible with Targeted Mass filter.
Remedy: Do not use MIPS Filter with Targeted Mass Filter.
214267
3 - Medium
Chromeleon: sporadic communication loss to Thermo Exploris Series ePanels.
Remedy: This may happen sporadically during standby or after a sequence has finished. Restarting Chromeleon allows regain control of Thermo Exploris Series ePanel.
233537
3 - Medium
Chromeleon periodically shuts down with a message “The target application domain has been unloaded.”
Remedy: Launch Chromeleon after shutdown. The sequence keeps on running without data loss.
357682
2 - High
Instrument Configuration: the “Wait for Temperatures to Stabilize checkbox” is not functional after an Instrument PC restart.
Remedy: Restart Instrument PC two times after enabling or disabling the configuration setting.
683124
3 - Medium
Customer/Tune: Favorites are not saved in the correct folder when upgrading from Exploris 4.2 SP4 to Exploris 4.3
Remedy: Copy the files from the Program Files location into the Program Data location.
758688
3 - Medium
Status Indicator doesn't turn red after Venting & Bakeout as supposed according fo Acceptance Criteria
828210
3 - Medium
System Calibration - IC Source calibration: Instructions panel (white box) shows incomplete feedback when procedure fails
843001
2- High
Max Injection Time: "Auto" does not take ETD overheads into account
844390
3 - Medium
Sequence stopped due to a commuincation issue between SII and Vanquish Neo
777178
2 - High
Sporadic DAQ USB misalignment errors during LC/MS sequence acquisition (Babble Error)