The table below lists new features, improvements and defect fixes in the Orbitrap Hybrid Series 5.1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 5.0 Instrument Control Software release.
Table Changes realized with Orbitrap Hybrid Series 5.1 Instrument Control Software relative to Orbitrap Exploris Series 5.0 Instrument Control Software
ID
Severity
Title
New features
Tune and Method Editor support new Orbitrap Exploris EFOX mass detector.
Improvements
842157
Inject Filtering: The instrument allows wider inject filtering for reduced fragmentation of intact proteins.
832170
OptaMax Plus: Vaporizer Temperature of 650 °C for the OptaMax Plus is available
851002
Mild trapping available on Exploris MX in Application Mode Small Molecule
Defect fixes
245291
2 - High
Chromeleon: Exploris Series ePanel shows disconnected but the instrument can still be controlled in Thermo MS Tuning, calibrations do not show in audit trail
332442
3 - Medium
Exploris GC: Self Calibration is turned on per default after upgrade from 4.1 to 4.2 (first time)
802783
3 - Medium
UI: ME: eDR: View Type: Quan View: Switch button is still visible
822716
2 - High
TNG Tune: 2nd graph discloses quadrupole plots that should only be shown with elevated rights
828499
3 - Medium
UI: ME: Activation Type - Compound specific settings - can be editable for not relevant rows
835457
3 - Medium
UI: ME: eDR m/z window tolerates first mass higher than 2500 for both eDR Window Types
835849
3 - Medium
UI: ME: Adaptive RT: not uploading a rtbin file with FAIMS if previously FAIMS voltage is not set to On
835858
3 - Medium
UI: ME: Adaptive RT alignment must be compatible with Negative polarity for MS2 scans
836578
3 - Medium
FAIMS: IC source cannot be run when FAIMS is installed on OE480 or Orbitrap Excedion Pro w/o ETD (SW error is thrown)
824387
3 - Medium
EThcD: Tune Spectrum View Scan filter displays obsolete values in free running mode
827917
2 - High
During the execution of sequences in Xcalibur, the following message may punctually be reported in warning popup: "Run preparation stopped with an error: GettingPreparedAgent receives Exception in waiting for the MS: The mass spectrometer doesn't assume the expected state."
844250
2 - High
Define Scan in Exploris MS tuning tab glitches to different settings in Chromeleon Console
856391
2 - High
Orbitrap Hybrid Series: MS sequences interrupting due to MS response timeout (partial fix)
The table below lists new features, improvements and defect fixes in the Orbitrap Hybrid Series 5.0 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.4 Instrument Control Software release.
Table Changes realized with Orbitrap Hybrid Series 5.0 Instrument Control Software relative to Orbitrap Exploris Series 4.4 Instrument Control Software
ID
Severity
Title
New features
396616
Orbitrap Excedion Pro - Method Editor: Adaptive RT Full Scan experiment enables Dynamic Time Scheduling for subsequent Targeted MS2 Scans, and also Data Dependent Scans using a Targeted Mass Inclusion Filter, without the need for spiking internal standards.
424869
Orbitrap Excedion Pro - Method Editor: Enhanced Dynamic Range (eDR) is available as new Full Scan property
351254
Orbitrap Excedion Pro - Method Editor: Pre-Accumulation for MS2 Scans is available as new Global Setting, and enables an additional Resolution 3750.
156646
Orbitrap Excedion Pro - Method Editor: Targeted Loss Trigger allows for triggering a dependent scan only when a specified loss ion is detected
746804
Orbitrap Excedion Pro BioPharma Edition provides an Extended Mass Range up to 12k Da for all supported scan types. Tune Calibration pane provides a new Calibration Type 'Extended Mass (AHFP)'.
785637
Orbitrap Excedion Pro supports the new Optamax Plus ion source
634919
Orbitrap Excedion Pro: Activation Types ETD and EThcD enable ETD (Electron Transfer Dissociation) to be optionally combined with supplemental HCD Activation (EThcD). The IC Source Calibration and Check are extended by ETD procedures.
775273
Orbitrap Excedion Pro: Hybrid-DIA - Adaptive RT tMS2 workflow supports DIA acquisition combined with Adaptive RT Dynamic Time Scheduling in one method.
155331
System Calibration: The System Calibration duration is shortened by an improved Inject Filter Isolation Calibration
Improvements
746032
FAIMS: PDF Reports contain additional Embedded System Information about the FAIMS configuration
776123
Method execution displays scan type in the RAW file scan header
789709
Tune: The "Smart burn-in of ICS" provides higher robustness and early stopping criteria, and replaces the "Short burn-in of ICS" procedure
Defect fixes
751317
3 - Medium
FAIMS: MS disconnection issues after running sequences in User Defined FAIMS Mode
337356
3 - Medium
Method Editor: Chromeleon does not apply method settings from System Templates when "keep current source settings" is selected
760961
3 - Medium
Method Editor: Targeted Mass Filter - Set Maximum Injection Time per Compound: The custom set value must be transferred to the Mass List table
769342
3 - Medium
Method Editor: Targeted Mass Filter with an empty Mass List Table may crash the Method Editor / is not functional when trying to clear or create a new method.
288280
3 - Medium
Method Editor: Template methods cannot be saved after saving a number of methods previously
692608
3 - Medium
Method Transfer: Erroneous Importing of Orbitrap Exploris 120 methods without AcquireX that were created with ICSW versions smaller than 3.0
826756
2 - High
Method Editor: Last mass for AIF isolation is too low (m/z 40-2500) for intact protein mode
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 4.4 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.3 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.4 Instrument Control Software relative to Orbitrap Exploris Series 4.3 Instrument Control Software
ID
Title
New features
440406
BioPharma: Intact Protein mode supports Full Profile mode
235125
Method Editor: Targeted Mass Filter supports maximum Injection Time as a table value for ddMS2 Scans
153450
Orbitrap Exploris 480 - TurboTMT: TMTpro reagents are expanded to 32-plex
Improvements
374516
Divert Valve: MS Valve turns to defined Waste Position in case of an MS instrument Error to protect the ion source from being flooded
455020
Exploris GC 240: "GCHigherResolution" license enables additional Resolutions 270 k and 300 k
458128
Exploris GC: Tune: Optimization tab provides new CI reagent gas flow optimization procedure
603775
Method Editor: Targeted Mass Exclusion filter is now enhanced by a toggle "Exclude isotopic cluster"
469661
Sequence acquisition stops when the instrument turns to "Source is open / Standby" mode
614368
System Calibration: New Calibration Type IC Source is now selectable separately
621042
System Calibration: Updated checkpoint settings are now permanently saved after each successful calibration step
306611
Tune: Alert Console emits an Error if UHV gauge is defective
Defect fixes
367365
Chromeleon: Method Editor does not size correctly in Chromeleon 7.3.2
749427
Chromeleon: Unable to create instrument methods with Chromeleon 7.3.2 MUc
757353
Exploris GC: Tune Optics are not loaded correctly
721841
FAIMS: MS disconnection issues after running sequences in User Defined FAIMS Mode
327877
Method Editor: Incorrect validation of hidden property "Scan Priority"
593860
Method Editor: Method execution of Decision Trees with Charge State dependent outcomes fails
378777
Method Execution Scan-to-Scan EASY-IC: In parts of the LC/MS run, the instrument does not detect the fluoranthene signal
753295
RAW file: Scan filter displays wrong average Collision Energies (CE) when doing multiplexed MS2 scans with stepped CEs
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 4.3 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.2 SP4 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.3 Instrument Control Software relative to Orbitrap Exploris Series 4.2 SP4 Instrument Control Software
ID
Title
New features
330867
Exploris GC: User-defined Lock Mass Correction Mode “RunStart” at the beginning of the data acquisition.
344696
Method Editor - Scan Properties: “Custom AGC Targets” display “Absolute AGC Values”.
349492
Method Editor - Scan Properties: “Use EASY-IC” is extended to Lock Mass Correction Mode “Timed”.
344714
Method Execution: extended the Maximum Injection Time Mode “Dynamic” to ddMS2 Scans.
344698
Tune and Method Editor OT Exploris 120 and MX: added Resolution settings 22500, 45000, 90000.
Improvements
321592
Exploris GC: Method Editor - Scan Properties: Product Ion Scan provides Product Ion Window Modes “Center Mass” and “m/z Range”.
326688
Exploris GC: Tune - Calibration: Manual Calibration table is separated by polarities.
455932
Exploris GC: Tune - Optimization: Leak Check has improved scan settings to eliminate m/z 42.
250676
Method Editor - Global MS Settings: “User-defined Lock Mass Injection” transitions to a Scan Property.
346290
Method Editor - Global MS Settings: Renamed “Internal Mass Calibration” to “Lock Mass Correction”.
340473
Method Execution: Instrument preserves the ion source properties defined in the instrument method after method execution has finished.
428795
OT Exploris 240: ddMS2/tMS2 scans allow for broader isolation windows up to 2000 m/z .
338482
OT Exploris MX: Tune Diagnostics - Define Scan functions are extended to Orbitrap Exploris MX to access all MALDI related functionalities.
297528
Tune - Alert Window emits a warning when starting Calibration/Check procedures while syringe pump is off.
277441
Tune Diagnostics - Tools allow for setting a PrOSA Limit value to exclude extreme PrOSA Scaling factors.
158424
Tune Preferences: Hotlink function is now limited to the Ion Source properties.
347290
Tune Preferences: The One-Point-Self-Calibration options are extended by a Calendar View.
358862
Tune: System Calibration stores all calibration values before the ICS Calibration, independent from the ICS HV Calibration outcome.
Defect fixes
381480
API: Using Handshake degrades instrument scan rate.
458619
Chromeleon - Exploris GC: Method Editor Throws TNG Errors in CM
360686
Chromeleon Studio is showing very slow performance, and sporadically no MS Device instrument.
390589
Ensure API test programs can load from path different to regular "programs" path.
443793
Exploris GC: Hotlink causes “Resolution” and "Scan Range Mode” cells flashing.
390653
FAIMS: instrument should be in standby while running Tune DV diagnostic procedure.
374578
Method Editor – DIA: error popup message “Object reference not set to an instance of an object” appears when modifying MSX ID values.
455541
Method Execution: EASY-IC on: Scan-to-scan cause interruption in rawdata file.
378777
Method Execution: In parts of the chromatogram the instrument does not detect the fluoranthene signal to correct the masses.
378826
Method Execution: unnecessary Hidden Scans/Polarity Switching in tMS2 Method with Funnel RF Level as Table Value.
437271
Method Transfer is not recognizing the OT Exploris model difference in Chromeleon [CM 7.2.10 MUh].
456234
Tune: sporadic false alerts “Source is open”.
485992
Sporadic missing scan in certain acquisition conditions (e.g., TurboTMT).
The table below lists defect fixes in the Orbitrap Exploris Series 4.2 SP4 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.2 SP3 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.2 SP4 Instrument Control Software relative to Orbitrap Exploris Series 4.2 SP3 Instrument Control Software
ID
Title
Defect fixes
424111
Software upgrade to version > 4.2 SP2 may result in a severe issue with loss of instrument configuration.
415092
After software upgrade from version 4.2 to a newer version, Tune displays obsolete warning alerts ([SSPI Link] – Reconnect during run-time) after firmware has been updated.
The table below lists defect fixes in the Orbitrap Exploris Series 4.2 SP3 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.2 SP2 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.2 SP3 Instrument Control Software relative to Orbitrap Exploris Series 4.2 SP2 Instrument Control Software
ID
Title
Defect fixes
356658
FAIMS Cooling Gas flow jumps to 20 L/min
404260
System Calibration: During Quad DC offset calibration, one point is sometimes too far off
405297
System Calibration: Quad isolation shape m/z 1034 iso 500 is not always detected
403256
System Calibration: Quad RF DDS frequency does not get set always on instrument boot
The table below lists defect fixes in the Orbitrap Exploris Series 4.2 SP2 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.2 SP1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.2 SP2 Instrument Control Software relative to Orbitrap Exploris Series 4.2 SP1 Instrument Control Software
ID
Title
Defect fixes
377461
System Calibration: C-Trap RF frequency calibration selects a frequency outside the optimum
384047
System Calibration: Orbitrap Exploris GC: C-trap RF Amplitude Min. Calibration failed with SW failure during run time
383676
Tune Status pane: Improved Turbo Pump readback robustness
The table below lists defect fixes in the Orbitrap Exploris Series 4.2 SP1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.2 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.2 SP1 Instrument Control Software relative to Orbitrap Exploris Series 4.2 Instrument Control Software
ID
Title
Defect fixes
356984
Method execution Full Scan DDA with Targeted Mass Filter and Fixed/Stepped Collision Energies (CEs): fixed CEs in the Targeted Mass Filter are erroneously executed as stepped CEs
347689
Method transfer: Cannot open certain older methods with Orbitrap Exploris Series 4.2 ICSW – index out of range error message
351444
Orbitrap Exploris GC: License Error with 60k GCHCD license prevents running methods
356173
Orbitrap Exploris GC: Method Editor: Corrected acquisition delay tooltip in Global Settings
356170
Orbitrap Exploris GC: Method Editor: Factor 10x UI recommendation rule is erroneously applied to all GC System Templates
356174
Orbitrap Exploris GC: Method Editor: Restore Default context menu does not work for parameters under Ion Source Properties
364748
Full Scan acquisition with Mild Trapping option (Small Molecule Application Mode) leads to signal drop in positive mode
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 4.2 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.2 Instrument Control Software relative to Orbitrap Exploris Series 4.1 Instrument Control Software
ID
Title
New features
153451
AcquireX support is provided for peptide mapping (AcquireX Ab) with Orbitrap Exploris 240 and Orbitrap Exploris 480
221915
Method Editor: New management of tables in DIA scan
168318
Method Editor: New table format is available for SIM scan. SIM with multiple broad scan ranges is provided for Orbitrap Exploris 120, 240 and GC.
153788
286667
Method Editor: Options are provided to select the order with which precursors are selected for data-dependent scans
186364
Method Editor: TMT 18-plex is supported by TurboTMT on Orbitrap Exploris 480
235118
Method Editor: The new option “Auto-Extended” is provided for MS2 Scan Range Mode (Small Molecule Application Mode)
285925
Orbitrap Exploris GC: Method Editor provides a “Run Start Mass Calibration” template in system template
290954
Orbitrap Exploris GC: Tune Calibration pane provides a Manual Calibration panel
284960
Orbitrap Exploris GC: Tune Preferences provides a panel for Self-Calibration Options
235073
Tune and Method Editor: Additional resolution settings are accessible for Orbitrap Exploris 240 and Orbitrap Exploris 480 (11.25k, 22.5k, and 90k)
214828
Tune: Scheduled one-point mass calibration (via Tune Preferences)
Improvements
302210
Diagnostics: FlexMix Spray Optimization (neg+pos) applies Source Gases independent from polarity
310820
FAIMS: Tune Calibration pane enables One-Point Mass and Customized Mass procedures which are compatible with FAIMS attached
208815
Method Editor: User experience is enhanced when changing “Max. number of multiplexed ions”
254731
Method Execution: Spray Voltage stays “On” during the execution of Run Start EASY IC
154537
Orbitrap Exploris GC: Chromeleon: Data Audit Trail exposes 'MS Tune Reports' from Thermo Exploris Series ePanel
Note This improvement is available only on Chromeleon versions ≥ 7.3.2.
318301
Orbitrap Exploris GC: Tune and ion source optimization reports contain a leak check history plot, filament current and emission current plots, and an emission current set value plot
327864
Tune and Method Editor: Optimized default ESI/HESI gas flow settings for Orbitrap Exploris 120, 240, MX
302209
Tune: Source gas and temperature are set independently from polarity
153260
Tune: Calibration Status panel: reworked the update of Recommended Calibration dates depending on the procedure outcome
308704
Tune: Calibration Status panel: clarified display of Last Successful Calibration to ‘Outdated’ after Venting and Bake-out
302209
Tune: Source gas and temperature are set independently from polarity
152982
Tune: Status pane: Added an exception for the “Temperature change since calibration” monitoring for automatically started bake-outs after venting the instrument
280624
The Orbitrap Exploris Series 4.2 Installer updates the MongoDB from version 4.06 to version 4.0.28.
Note MongoDB needs to be uninstalled manually when downgrading from version 4.2 to an older version. The ISO image contains a MongoDB uninstaller.
Defect fixes
291191
AcquireX: Method Editor needs to mark incompatible Mass List Types in red
203732
Chromeleon: Method Editor menu bar is missing
302180
Method Editor / Instrument Configuration: Settings “Contact Closure” are named the same but refer to different functions (Digital Input/Output)
245745
Method Editor: MSX ID in Mass List Table changes when switching between Quan and Qual Views
221534
MongoDB uses too large memory (>3GB) at background
318660
TurboTMT: Method Editor needs to mark the incompatible MS2 property “Scan Range Mode = Auto” in red
329295
Workstation method editor cannot be launched without Foundation installed (affects Chromeleon Enterprise)
322914
Installation of workstation OES removes previously installed workstation version
297887
System Calibration: Electronics Smart Calibration is checking against the wrong frequency range in interference peak removal procedure
The table below lists improvements and defect fixes in the Orbitrap Exploris Series 4.1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.0 SP1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.1 Instrument Control Software relative to Orbitrap Exploris Series 4.0 SP1 Instrument Control Software
ID
Title
Improvements
270859
FAIMS Pro and Pro Duo: Adjusted fan speed monitoring thresholds
254371
FAIMS Pro and Pro Duo: DV Tune and DV Check procedures operate at a wider DAC range to support FAIMS operation at altitudes > 2000 m
263849
FAIMS Pro and Pro Duo: Improved gas control to conserve cooling gas
218222
Instrument Manuals: PDF documents provide translations of the English Operating Manual and Pre-Installation Requirements Guide in Chinese, French, German (Operating Manual only), Italian, Japanese, and Spanish
229906
Tune System Calibration includes a reworked noise-canceling procedure (Interference Peak Removal) as part of the Electronics Smart Calibrations
154508
Tune System Calibration runs independent from selected Application Modes (Small Molecules, Peptide, Intact Protein)
250289
Tune System Calibration/Check: Improved robustness of the HCD Fragmentation and Trapping Efficiency Check
242936
Tune System Calibration: Further improvements of the Isolation Shape Analysis algorithm to obtain a shorter duration of system calibration procedure
232380
Tune System Calibration: HCD Fragmentation and Trapping Efficiency Check is now also part of the System Calibration
243124
Tune: The About Tune box now shows active licenses for all Orbitrap Exploris mo
156731
Tune: The Status pane (By Function / By Board) displays Turbopump readbacks in the Vacuum System node
152982
Tune: The Status pane (By Function) indicates the ambient temperature change since last calibration in the new Performance node
633
Orbitrap Exploris GC: Apply a yellow warning when a fullscan range is beyond the 15x rule in method editor
622
Orbitrap Exploris GC: Remember the last used polarity to an added new line in the internal lock mass table in method editor
653
Orbitrap Exploris GC: Enable MSMS and all resolutions on the Standalone Workstation
Defect fixes
164799
Chromeleon: The instrument method of extracted raw files now shows the Orbitrap Exploris model as instrument name
245000
Chromeleon: Method Editor shows AcquireX UI Elements when instrument is operated by Chromeleon
153267
Chromeleon: Method Editor: Cannot edit Mass List Tables when maximized as popup window
267594
Method Editor: Added 45000 resolution for DIA Scans
233648
Method Editor (Full Installation): RF Level Conversion is erroneous when importing methods between Orbitrap Exploris 120/240/MX and Orbitrap Exploris 480
258112
Method Editor Ion Source Properties: Gas Mode settings are available when Source Type is NSI
243767
Method Editor: Orbitrap Exploris 120 Mass List Tables: MSX-IDs are changed when saving and reopening the method
263755
Method Editor: tMS2 Scan: Method Download Validation fails with stepped Collision Energy
280344
Method Editor: When “RunStart” is selected in the global MS Settings, scan properties show “Use EASY-IC” parameter
258279
Method Execution: Multiplexing in tMS2 negative mode is not executed when polarity is a table value
261532
Tune Calibration Status: Recommended Calibration Dates need to reset after Venting and Bake-out event
258076
Thermo Foundation Instrument Configuration: The Contact Closure configuration dropdown list labels are now aligned with the Method Editor (Transition close-to-open, Transition open-to-close)
258413
Nanospray Flex Ion Source: Source interlock does not always prohibit acquisition mode
625
Orbitrap Exploris GC: Correct “Collision Energy Type” in MS2 Scan type in Tune
627
Orbitrap Exploris GC: Correct “HCD collision energy” tooltips in Method Editor
632
Orbitrap Exploris GC: Correct “Negative Quad Isolation Check” error message
636
Orbitrap Exploris GC: Add “Ion Source Optimization Report” Header to the optimization report in Tune
637
Orbitrap Exploris GC: Default “Hot link” is unchecked in Tune Preferences
640
Orbitrap Exploris GC: Change default mass and mass range under Optimization in Tune
661
Orbitrap Exploris GC: Correct “Injection Flatapole DC and Inter Flatapole Lens” names that associate to the corresponding procedures in Tune
208147
Orbitrap Exploris GC: Plot Chromatogram is not active when the system is on
250789
Orbitrap Exploris GC: User saved “Favorites” settings in Tune are not loaded if MS2 scan is defined with normalized collision energy
265908
Tune Diagnostics: Removed “ICD Calibration (No Ions Required)” from Partial Calibration tree
153053
Tune Plots: All plots need to clear when going to the next sub procedure
154440
Tune Plots: Context menu “Copy to Clipboard” needs to be functional outside the graph plot area
276236
TNG Tune: Set Value Positive Ion Spray Voltage shows value “0” instead of actual set value when running sequences
The table below lists defect fixes in the Orbitrap Exploris Series 4.0 SP1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 4.0 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.0 SP1 Instrument Control Software relative to Orbitrap Exploris Series 4.0 Instrument Control Software
ID
Title
Defect fixes
252969
System Calibration: eFT calibration may fail due to wrong fit
253113
Orbitrap Exploris MX System Check: Negative HCD Fragmentation and Trapping Efficiency Check may fail on too low m/z 1506 intensity
The table below lists new feature, improvement and defect fixes in the Orbitrap Exploris Series 4.0 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 3.1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 4.0 Instrument Control Software relative to Orbitrap Exploris Series 3.1 Instrument Control Software
ID
Title
New features
188489
Tune: Customized Mass Calibration and Check run the Mass Calibration with User-definable mass lists.
190826
Tune: Calibration pane provides a One-Point Mass Calibration procedure using fluoranthene.
152876
Tune: Fore Vacuum supervision: The user needs to be informed if transfer tube is clogged.
153034
Method Editor: Global Settings - Internal Mass Calibration provides various internal calibration modes (RunStart, Scan-to-Scan, Timed).
154268
Method Editor: Global Settings - Internal Mass Calibration offers Lock Mass Injection functionality for user-defined lock masses.
153449
Raw file spectra provide access to extended peak data supported by FreeStyle 1.8 and higher
Improvements
235608
Tune and Method Editor Precursor m/z: Quadrupole Isolation Range checking needs to consider the first mass of the quadrupole isolation window.
236947
Tune and Method Editor Scan Range checking needs to restrict Full Scan ranges to the Factor 15 rule (Orbitrap Exploris 120, 240, 480, and MX).
214822
Tune Diagnostics: rearranged the Tools tree for improved access to the toggles and settings separated by category: Calibration, Define Scan, Application Mode, Method Setup, Peripherals.
193411
Tune System Calibration: Faster, more robust Isolation Shape Analysis algorithm results in shorter duration of system calibration procedure.
557
Tune: Mass Calibration Due Time is active in Diagnostics Pane also for Orbitrap Exploris GC.
629
Orbitrap Exploris GC Tune: Move “Quadrupole Isolation Check” and “Quadrupole Calibration” to Diagnostics Pane.
604
Orbitrap Exploris GC Tune: improvements in the Quad Iso Calibration procedure.
195617
Chromeleon: ePanel Thermo MS Tuning: Instrument Audit Trail logs information about Tune events.
225995
FAIMS Pro Duo: a firmware update mitigates 'overcooling' of the FAIMS electrodes.
214725
Software Manuals / Online help: New website https://www.analyteguru.com/ succeeds deprecated PlanetOrbitrap.com.
156632
Thermo Foundation Instrument Configuration: Ion Source: When toggle “Enable Sweep Gas for NSI Source” is disabled, the sweep gas will disappear from the Tune and Method Editor user interface, and inactivate the sweep gas flow.
243744
Method Editor: Full Scan: added a recommendation to set the first mass to values ≤2500 m/z.
203331
Method Editor: Targeted Mass Filter: Property 'Collision Energy Mode' is removed and HCD Collision Energy (% or V) allows access to 1 (fixed), and 2, 3, 4, 5 (stepped) set values.
211730
Method Editor: Targeted Scans: Property 'Collision Energy Mode' is removed and HCD Collision Energy (% or V) allows access to 1 (fixed), and 2, 3, 4, 5 (stepped) set values.
559
Orbitrap Exploris GC Method Editor: Add property “Isolation offset” to SIM scan.
217910
Method Execution: EASY-IC applies the same lock mass correction behavior for ddMS2 scans as with User Defined Lock masses.
213667
Orbitrap Exploris 120 provides access to precursor m/z isolations up to m/z 2500 in ddMS2/tMS2 and SIM scan types.
Defect fixes
204120
Tune Diagnostic procedure for ICS Burn-in and HV Current Calibration unexpectedly requires FlexMix infusion.
533
Orbitrap Exploris GC Tune: Limited the mass range in plot chromatogram and monitor mass accuracy tables.
555
Orbitrap Exploris GC: Reorder repeller and source offset values recorded in raw files under Optics table of Instrument Method section.
153327
Chromeleon: AcquireX UI Elements show in Method Editor when instrument is operated by Chromeleon.
233966
FAIMS and tSIM ddMS2 - Method Execution: MS2 scans are collected at the same CV, regardless of the CV used as a table value in the master SIM scan.
245244
FAIMS: Tune Ion Source pane: Sweep Gas is set when using “Get Default” function.
153573
Thermo Foundation Instrument Configuration: Behavior of “Enable Sweep Gas for NSI” differs from Tribrid Series Instrument Control Software.
235015
Method Editor: 45000 resolution is missing for Full MS and SIM.
153360
Method Editor: Ion Source Properties: UI validation formula for gas flows is incorrect.
243792
Method Editor: Mass List Tables: Within the same User-defined MSX ID group, different HCD Collision Energies and Adducts are restricted erroneously.
243763
Method Editor: Mass List Tables: Collision Energies (CE) are not imported when CE types (Absolute, Normalized) are different.
218273
Method Editor: tSIM + MSX methods with adjacent mass ranges are correctly validated but not executed.
237786
Method Execution: EASY-IC does not correct masses in ddMS2 scans.
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 3.1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 3.0 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 3.1 Instrument Control Software relative to Orbitrap Exploris Series 3.0 Instrument Control Software
ID
Title
New features
153032
FAIMS Pro Duo Interface: Tune and Method Editor support Highflow LC/MS applications using HESI/APCI sources
152894
Tune: Calibration pane user interface is aligned with Tribrid Series 3.4
186434
Orbitrap Exploris 120 Method Editor provides more options: up to five experiments per method, Microscans, Multiplexing of up to 20 targets, Custom AGC Target
174361
Method Editor provides an Isotope Exclusion Filter to choose assigned or unassigned isotopes for triggering data dependent scans
Improvements
190370
FAIMS Pro Duo: improved AGC performance when method applies different Compensation Voltages (CV)
199341
FAIMS Pro Duo: High resolution mode definition: inner electrode temperature increased from 70 °C to 80 °C
156627
Data acquisition: Intensity display correction for very low precursor isolation widths
155669
Tune: System Calibration: improved resume function from the last valid checkpoint if procedure has failed
194014
Tune: System Calibration: improved Injection Filter Low-m/z RF Calibration to enhance ion transmission for m/z values < m/z 150 on some systems
209230
Tune: Calibration option 'Skip Spray Stability Evaluation for Mass Calibration' moved to the Diagnostics pane
189067
Tune and Method Editor support MALDI and DART sources for Orbitrap Exploris 120, 240, 480
202700
Tune: improved readability of PDF Reports for calibration, check, and diagnostics procedures
174997
Method Editor: Targeted MS2 scans and Targeted SIM scans support Polarity as a table value also when multiplexing in tSIM and tMS2 experiments
153453
Method Editor: Targeted MS2 scans support Fixed and Stepped Collision Energies as a table value also in multiplexing tMS2 experiments
166860
Method Editor: Targeted Mass Filter supports Stepped Collision Energy
187465
Method Editor: RunStart EASY-IC scans are moved to the “Prepare for Run”-time and consider more than the very last lock mass correction for a more reliable lock mass correction at the beginning of the data acquisition
155244
Method Editor: Mass List Table UI recommendations are aligned with Tribrid Series 3.4
175638
Orbitrap Exploris 480 Method Editor: Dynamic Exclusion Filter supports prefiltering data dependent scans with multiple branches
Defect fixes
201250
Data Acquisition: Missing masses in Full MS when first mass is set > m/z 2500
202960
FAIMS Pro Duo: DV Tune procedure gives temporary false error alert
177971
Tune: Calibration: System always switches to Low Pressure after procedure run if in Intact Protein mode regardless of prior pressure
204120
Tune: Diagnostic procedure for ICS Burn-in and HV Current Calibration unexpectedly requires FlexMix infusion
152898
Tune: Error message for HESI vaporizer readback is incorrect on certain instruments due to wrong delta value
153397
Tune: System Calibration: When calibration fails, Recommended Date info gets lost and Performance Status shows an Error
183927
Handshake: When recording a RAW file triggered by handshake mode, one extra scan is added into RAW file
190496
Method Editor Summary: Error when printing (complex) instrument methods
198382
Method Editor: DIA method does not start because calculated window lies outside precursor mass range when Window Placement Option is active
207283
Method Execution Orbitrap Exploris 480: Cycle time is longer than defined in the method when multiple ddMS2 branches use scan priority
546
Orbitrap Exploris GC - Tune: Leak Check fails when Scans to Average is turned on in Tune.
540
Orbitrap Exploris GC - Tune: Incorrect Theoretical Masses for Negative Ions are shown in Customize Calibration within Tune.
541
Orbitrap Exploris GC - Tune: Users cannot run a normal mass calibration after a custom calibration. Customize Calibration runs even if it is not selected.
186
Orbitrap Exploris GC - Tune: Selecting “Abort” does not stop all selected Ion Source Optimizations.
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 3.0 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 2.0 SP2 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 3.0 Instrument Control Software relative to Orbitrap Exploris Series 2.0 SP2 Instrument Control Software
ID
Title
New features
152951
FAIMS Pro: Data acquisition optimizes FAIMS CV post-switching delay time when using multiple CV Voltages per experiment
153796
FAIMS Pro: Total Carrier Gas Flow succeeds FAIMS User Gas to enable lower flow rates for improved nano spray (NSI) stability
153696
FAIMS Pro: Instrument configuration supports user-defined IP addresses for FAIMS
152892
Tune Diagnostics provides restructured procedures separated by category (Partial Calibration, Optional Calibration) and polarity (Positive, Negative)
152877
Tune Diagnostics provides Low m/z Spectral Mass Accuracy Calibration and Check to obtain high mass accuracy also in the low mass range
154453
Tune Diagnostics provides a tool to change the Mass Calibration Due Time
153319
Tune supports Collaborator interfaces via license
153450
Method Editor supports TMTpro Reagent (TMT-16-plex) for Orbitrap Exploris 480
163618
Method Editor supports variable collision energies in a Mass List Table for multiplexing experiments
Improvements
153497
Tune: Instrument Status pane displays vacuum pressures in a separate heading “Vacuum System”
152895
Tune: System Calibration and Check stop the syringe pump when procedure has finished to save consumption of FlexMix
152998
Tune: System Calibration: Calibration Solution Pre-Check forces the ion transfer tube to 320 °C for optimal spectral quality of FlexMix
152997
Tune: System Calibration includes an Interference Peak Removal procedure (only run after Venting and Bake-out)
152996
Tune: System Calibration provides an improved Quadrupole Main and End Segment DC Calibration
185621
System Calibration Negative: Improved robustness of ICS Active HV Current calibration subprocedure so that the procedure runs successfully after the first attempt
152994
Tune: System Check checks HCD Fragmentation/Trapping in both ion polarities
152978
Tune Diagnostics provide Tooltips for the whole diagnostics procedure tree
152897
Tune Diagnostics provide High Mass Range Calibrations (AHFP) also for Negative polarity when using Application Mode 'Intact Protein'
153051
Tune Diagnostics provides an improved Quadrupole Maintenance Check procedure
152983
Tune Diagnostics provides an improved Transfer Tube Maintenance Check procedure
154542
Tune Diagnostics procedure “Disable AGC” disables PrOSA when AGC mode = fixed
155647
Help Files: Method Editor provides an Online Help
155646
Help Files: Tune provides an Online Help
164760
AcquireX: Improved robustness for running experiments with Mass List Tables >5000 entries
152995
BioPharma: Improved eFT phase calibration to improve mass accuracy for High Mass Range applications
173884
tSIM Isolation Offset: the user can define an isolation offset for the isolation windows. The parameter is accessible as table value
152874
Method Editor: user needs to access updated System Templates as applies with the upcoming release OES 3.0
152875
Method Editor: Global MS Settings provide optimized default values per selected Application Mode (Small Molecule, Peptide, Intact Protein)
152979
Data Acquisition: RAW files include signal instability warnings in the Error Log
Defect fixes
152969
Chromeleon: User cannot open Method Editor Workstation Standalone from client computer
153344
Data Acquisition: Instrument still scanning in Standby and Off mode
183927
Data Acquisition: When recording a RAW file triggered by handshake mode, one extra scan is added into RAW file
153308
EASY-IC: In Polarity Switching experiments, ICS AGC is not done for each polarity separately
166232
Method Editor: DIA peak lists do not give original values when Mass Defect is set back to 1.0005 m/z
180523
Method Editor: Could not run Quan View DIA methods when Precursor Mass Range exceeded last mass m/z 524
166862
Method Editor: DIA scan Window Placement Optimization not properly applied for non-zero Window Overlap value
153270
Method Editor: Incorrect RF Lens Value Shown when Importing Methods between Orbitrap Exploris models
152940
Method Editor: Targeted Mass Filter does not support Fixed Collision Energy as table value (SureQuant)
170714
Method Execution: Dynamic exclusion filter: Mass tolerance mode “m/z” not working
153420
Method Validation: Mismatch of Ion Source type configuration during sequence submission
153577
Tune: System Calibration: Not all 'after venting procedures' run after venting - and 'after venting procedures' run although no venting took place
162574
Tune: Ion Source pane calls wrong default (H)ESI settings when 'Current LC Flow' = 500 μL/min
The table below lists improvements and defect fixes in the Orbitrap Exploris Series 2.0 SP2 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 2.0 SP1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 2.0 SP2 Instrument Control Software relative to Orbitrap Exploris Series 2.0 SP1 Instrument Control Software
ID
Title
Improvements
9783
Orbitrap Exploris 120 Method Editor: Extend Data Dependent MS2 Method to create TopN experiments with N=1,2,3,4 in DDA
9781
Orbitrap Exploris 120 Method Editor: Provide System Templates for TopN experiments with N=1,2,3,4
Defect fixes
9667
BioPharma: Application Mode "Intact Protein" uses unsafe Extended Trapping 1 eV
9849
EASY-IC: ICS Prechecks should give dedicated error for extreme mass deviation
9683
EASY-IC: Issues with ICS ion intensities after prolonged idle time
9817
RunStart EASY-IC: Miscorrection when polarity switching is active
9844
System Calibration: C-Trap RF Frequency Calibration should avoid 3.2 MHz
The table below lists improvements and defect fixes in the Orbitrap Exploris Series 2.0 SP1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 2.0 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 2.0 SP1 Instrument Control Software relative to Orbitrap Exploris Series 2.0 Instrument Control Software
ID
Title
Improvements
9409
Limit maximum C-Trap RF for increased robustness
9261
BioPharma: Provide AHFP calibration in TNG Tune Diagnostics
Defect fixes
9228
BioPharma Orbitrap Exploris 240: Loss of signal at maximum S-lens RF amplitude during intact mAb LC-MS analysis
9498
DIA: Method transfer from Orbitrap Exploris Series ≤ 1.1 SP1 to Orbitrap Exploris Series 2.0 does not consider the new AGC Target Normalized Base 1e5
9267
Diagnostics: Quadrupole Maintenance Test switches rod configuration but does not apply rod-specific calibration set
9524
FAIMS communication LED remains off when ion source is removed and reconnected
9242
FAIMS DV Tune and Check are not always shown in Tune Diagnostics pane
9461
FAIMS: In-source CID together with FAIMS results in a defect raw file
9464
Method Execution: In-source CID is not applied for data-dependent scans
9341
System Calibration: Checkpoint “Inject Filter (IF) Calibration” separates IF Isolation Calibration and Check, trapping users with poor IF calibrations
9470
UI: Method Editor crashes when deleting Targeted Mass Exclusion Filter from the method
9283
UI: Tune: Ion Source tab: “Get Defaults” ion source values for “Current LC flow” does not work
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 2.0 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 1.1 SP1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 2.0 Instrument Control Software relative to Orbitrap Exploris Series 1.1 SP1 Instrument Control Software
ID
Title
New features
4846
AcquireX: Orbitrap Exploris 240 and 480 support AcquireX workflows for Application Mode “Small molecule”
765
iAPI provides access to real-time control of Orbitrap Exploris 480
7514
Method Editor provides access to latest System Templates updates directly via the Thermo Fisher Cloud
8144
Method Editor: Application Mode ‘Small Molecule’ provides optionally access to Mild Trapping settings to reduce MS1 fragmentation of labile compounds
7513
Method Editor: Ion Source pane provides access to time-dependent ion source gas flows
5121
Method Editor: RunStart EASY-IC allows for internal mass calibration only at the beginning of the data acquisition
7512
Method Editor: tMS2 and DIA scans use isochronous injection times for multiplexing experiments
322
Method transfer: Method Editor is able to open and save methods from all Orbitrap Exploris Series instruments
7953
Tune Calibration pane: System Calibration provides improved help text in gray box, Alert Window and Calibration Reports
8174
Chromeleon: Orbitrap Exploris Series 2.0 shall run under Chromeleon 7.2.10
8173
Chromeleon: Orbitrap Exploris Series 2.0 shall run under Chromeleon 7.3
Improvements
5012
EASY-IC: Method Execution and Tune operate IC Source in idle mode when EASY-IC is not use
8090
EASY-IC: Thermo Foundation Instrument Configuration provides the possibility to switch off the IC Source
7545
FAIMS Pro provides same optimized CV values aligned with Tribrid Series and TSQ Series instruments
7121
Method Editor: DIA Scan AGC target is aligned with Tribrid Series - Normalized Base (100%) is equal to 1e5
8542
Method Editor: Mass List Tables: MRFA is default compound aligned with Tribrid Series
8816
Method Editor: Mass List Tables: Pop-up Warning 'Columns don't match' has improved messages when importing Exactive Series Global Lists
7119
Method Editor: Targeted MS2 Scans: 'Dynamic RT' provides a Retention Time Standards Table for Pierce PRTC Mixture (Orbitrap Exploris 240 and Orbitrap Exploris 480 models)
7410
Scan Filter: Methods using Internal Mass Calibration hide “lock” to improve generating XICs from the same scan filter
8001
Tune Calibration pane: Recommended Date of a mass calibration updates after successful system calibration
643
Tune: Ion Source pane provides improved default Ion Source Parameters
Defect fixes
7919
Installer: Uninstallation report of Orbitrap Exploris Series 1.1 shows content of installation
7840
Instrument stays in Off mode after Data System Computer Reset
6605
Method Editor: Audit trail shows wrong number of data dependent scans
8747
Method Execution: Too much precursor ion intensity is remaining when using stepped (N)CE
8555
RAW file Scan Header: Precursor Fit Filter: 'Error in isotopic envelope fit' is not recorded
4972
RAW files: Instrument Method settings do not reflect Method Editor Summary content
8489
Tune cannot start because TNGConfig.xmb file gets corrupted sporadically
6095
Tune Diagnostics pane shows FAIMS procedures although FAIMS is not attached with the instrument
8869
Tune: Application Mode 'Intact Protein': Trapping voltage of Continuous acquisition is misaligned with Instrument Method acquisition
The table below lists improvements and defect fixes in the Orbitrap Exploris Series 1.1 SP1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 1.1 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 1.1 SP1 Instrument Control Software relative to Orbitrap Exploris Series 1.1 Instrument Control Software
ID
Title
Improvements
8053
Instrument Configuration: Check box “Enable Minimum Gas Flow Requirement for Ion Source” replaces the previous text field.
8348
System Calibration: Improved Bent Flatapole RF calibration.
8162
System Calibration: Improved Quad Transm./Rod Config Calibration.
8115
System Calibration: Removed checkpoint after quadrupole isolation calibration.
Defect fixes
8128
Alerts Console: Low OT temperature readback raises “Temperature too high” failure.
8297
Chromeleon: MS Tuning ePanel: Passed calibration file is named as Failed.
8180
Instrument Status: Electronics does not recognize readbacks being outside possible set range.
8284
Instrument Status: Threshold for warning “UHV gauge not ignited” is too high.
8070
Method Editor: System Templates: SureQuant method is not fully functional.
8031
Method Editor: TopN with multiple outcomes and ddSIM: Method Editor closes after appending a Targeted Mass Filter.
8040
System Calibration: Wrong threshold used in IF Low m/z RF Calibration.
The table below lists new features, improvements and defect fixes in the Orbitrap Exploris Series 1.1 Instrument Control Software release that were implemented since the Orbitrap Exploris Series 1.0 Instrument Control Software release.
Table Changes realized with Orbitrap Exploris Series 1.1 Instrument Control Software relative to Orbitrap Exploris Series 1.0 Instrument Control Software
ID
Title
New features
7538
System Calibrations must provide improved user experience.
User Story 4140—Tune: Diagnostics provide single-component sub procedures of system calibration.
User Story 6961—Tune Calibration: System calibrations are resume-able. Intermediate results are saved as checkpoints. This allows for optimizing spray conditions or refilling the syringe.
365
The user must be able to define favorite scan settings for easy access to sets of parameters.
User Story 748—Tune: Favorites pane: System Settings: Favorite settings for ESI source provide default ion source and define scan properties for calibration and diagnostics procedures.
7084
The Dynamic Exclusion filter must support a global list option (exclusion across experiments and outcomes).
User Story 819—Method Editor: Dynamic Exclusion Filter provides global functionality across all experiments in the method time line (new property “Share dynamic exclusion list with other selected dynamic exclusion filters”).
7126
Method Editor: Must have import list functionality from QE exported lists.
User Story 7339—Method Editor: Import of Exactive Series mass lists (CSV files) into Orbitrap Exploris Series 1.1 mass list tables is supported.
  • Exactive Series global inclusion lists can be imported into Orbitrap Exploris Series mass list tables of scans (tMS2, Product Ion Scan, SIM Scan) and filters (Targeted Mass Filter).
  • Exactive Series global exclusion lists can be imported into Orbitrap Exploris Series Targeted Mass Exclusion Filter.
762
The instruments must be able to interface with Chromeleon Software.
User Story 3174—Chromeleon: Tune and Method Editor are integrated in Chromeleon 7.2.10 (please install Chromeleon 7.2.10 hotfix files from thermo.flexnetoperations.com or from support.thermoinformatics.com).
Improvements
7297
FAIMS: Acquisition sequence will pause if system recognizes a DV drop. This gives the opportunity to run a DV Check or DV Tune procedure before resuming the sequence.
7538
System Calibrations must provide improved user experience.
User Story 6227—Tune Diagnostics: improved bake-out procedure.
User Story 6228—Tune Calibration: System calibrations are improved regarding robustness.
User Story 7167—Tune Diagnostics: improved ICS calibrations (EASY-IC option).
User Story 7295—Tune Calibration: Additional calibration information is displayed in gray box.
User Story 7328—Tune Diagnostics: “FlexMix Solution Evaluation” procedure to check for contaminated FlexMix.
7231
Tune Diagnostics: improved ion optics maintenance check.
7587
Tune Alert Window: improved messages about instrument status, calibration/check and diagnostics.
371
Users must have access to user manuals and online Help.
User Story 2145—UI: Tune shall display tooltips for each UI element.
User Story 2146—UI: Method Editor shall display Tooltips for each UI element.
7098
Updated system templates must be applied.
User Story 7420—Method Editor: Templates tab provides updated system templates for all application modes and for all models of Orbitrap Exploris Series MS systems.
Defect fixes
6490
Tune: APCI Source Optimization: Pos and Neg ion discharge current not supported.
6493
Tune: APPI Lamp of APPI ion source not controlled.
6733
Chromeleon: Data Acquisition via MS Tune panel is not supported.
6736
Chromeleon: Instrument Status ePanel needs to be configured manually.
6760
Chromeleon: Anomaly on first sequence acquisition in a series of sequences.
6790
Chromeleon: User mode selection anomaly.
6799
Instrument needs to be set to standby manually after reboot.
6812
ME and Tune GUI allows isolation low being higher than 2500.
6817
User does not reliably get a message when vacuum is not ready for instrument electronics to be set to “On” or “Standby.”
6819
Tune: Define Scan: Collision Energy Mode: Stepped CE is not supported.
6821
PDF Reports: Diagnostics: Quadrupole Maintenance Check Plot not included.
6850
Tune: Status Pane: Peripheral Devices: “Turbo Pump 1” show Status “Not Connected” although turbo pump is connected.
6877
Data Acquisition via Method: Source type “ESI” not supported.
6929
After boot, Orbitrap Supply throws a warning (Device Limit exceed and Status-Warn), which can be ignored.
6930
Sometimes system calibration can fail due to unstable spray.