The minimum hardware and software configurations required for Orbitrap Hybrid Series 5.1 Instrument Control Software operation are as follows:
System | Requirements |
|---|---|
PC | 3.0 GHz Quad Core Intel™ Processor 32 GB RAM 512 GB SSD Hard Drive Display Monitor Resolution of 1920 × 1080 Two Network Interface Cards (NIC), 1000 MBit/s |
Software |
|
TIP
The Orbitrap Hybrid Series 5.1 Instrument Control Software was only tested within the delivered composition.
Support for Chromeleon™. The Chromeleon driver compatibility matrix reflects the tested compatibility for combinations of Orbitrap Exploris Series instrument control software versions, Chromeleon software versions, and instrument models. For more information and to view the compatibility matrix, sign on to support.thermoinformatics.com/downloads/default.aspx, and then select Chromeleon > Chromeleon > Related Drivers > Driver Compatibility Matrix.