The following table summarizes which filter types are available to append to which scan types.
Filter Types
Scan Types
MS
tSIM
tMS2 / tMS3
tMSn
ddSIM
ddMS2
ddMSn
Precursor Selection Range
ü
ü
ü
ü
ü
ü
MIPS
ü
ü
ü
ü
ü
ü
Intensity
ü
ü
ü
ü
ü
ü
Precursor Fit
ü
ü
ü
ü
ü
ü
Charge State
ü
ü
ü
ü
ü
ü
Dynamic Exclusion
ü
ü
ü
ü
ü
ü
Isotope Exclusion
ü
ü
ü
ü
ü
ü
Real-Time Library Search filter
ü
ü
Targeted Inclusion
Targeted Mass
ü
ü
ü
ü
ü
ü
Targeted Mass Difference
ü
ü
ü
ü
ü
ü
Targeted Isotopic Ratio
ü
ü
ü
ü
ü
ü
Targeted Loss Inclusion
ü
ü
ü
Targeted Exclusion
Targeted Mass Exclusion
ü
ü
ü
ü
ü
ü
Precursor Ion Exclusion
ü
ü
ü
Targeted Loss Exclusion
ü
ü
ü
Apex Detection
3
Triggers
Targeted Mass Trigger
ü
ü
ü
ü
ü
ü
Targeted Loss Trigger
ü
ü
ü
MSn Quality Trigger
ü
ü
ü
Alternate Precursor Sorts
Sort by Intensity
ü
ü
ü
ü
ü
Sort by m/z
ü
ü
ü
ü
ü