The Orbitrap Tribrid Series instruments offer a range of optional licenses and add-on options. Some options require factory installation at the time of purchase, while others are available later as field upgrades.
License | Description |
|---|---|
IRMPD | Factory-installed option available only at the time of purchase; not available as a field upgrade. Includes the IR laser module and a license that enables the infrared multiphoton dissociation (IRMPD) activation type. NOTE: IRMPD includes corresponding hardware modifications to support the software features. |