Welcome to the Orbitrap Tribrid Apex Small Molecule Method Editor Help for the Thermo Scientific™ Orbitrap™ Tribrid™ Series 5.0 Instrument Control Software. This Help system describes how to use the Method Editor application with the mass spectrometer (MS). You create an MS instrument method (combined with the optional autosampler and liquid chromatograph instrument methods) by defining the experiment type and by setting various parameters. This includes settings for the MS, syringe pump, and divert valve; and the mass ranges and fragmentation transitions for the experiments.
See the following introductory topics: