The following tables list the accepted values between the Quadrupole and Ion Trap isolation modes.
The validation rules applied for tSIM and tMS2 scans now considers the entire m/z range to be isolated. This range is a combination of the target m/z (center mass) and the isolation window. To pass validation, the software considers the isolation range, center mass, last mass, and first mass. Each of these parameters is considered in series.
License | Scan Type | Detector | Isolation Mode | Last Mass | First Mass | Isolation Width |
|---|---|---|---|---|---|---|
Standard | tSIM | Orbitrap | Quadrupole | 50–2,000 | 50–2,000 | 0.4–2,000 |
Ion Trap | Quadrupole | 50–2,000 | 50–2,000 | 0.4–2,000 | ||
tMS2 | Orbitrap | Quadrupole | 40–2,000 | 40–2,000 | 0.4–5 | |
Ion Trap | Quadrupole | 40–2,000 | 40–2,000 | 0.4–5 |
License | Scan Type | Detector | Isolation Mode | Last Mass | First Mass | Isolation Width |
|---|---|---|---|---|---|---|
Standard | tSIM | Orbitrap | Ion Trap | 50–2,000 | 50–2,000 | 0.2–2,000 |
Ion Trap | Ion Trap | 50–2,000 | 50–2,000 | 0.2–2,000 | ||
tMS2 | Orbitrap | Ion Trap | 40–2,000 | 40–2,000 | 0.2–5 | |
Ion Trap | Ion Trap | 40–2,000 | 40–2,000 | 0.2–5 |