The following table describes the parameters in the Preferences
area.
Parameter | Description |
|---|---|
Enable Recommendations | Shows (or hides) tooltips that specify recommended values or ranges. Default: Selected When you enter a value or range that is not recommended or outside the recommended range, the Method Editor application changes the border color of the affected workflow object and parameter box to yellow. Do the following:
|
Enable Favorites | Shows (or hides) the stars that designate favorite parameters. Default: Selected |
Restore Default Favorites | Resets all favorites to the default factory setting. Default: Deselected |
Restore Advanced Template Messages | Restores the System Templates dialog box that prompts you when a template has additional online information. Default: Deselected |