Select the Output Contact Closure checkbox to open the Output Contact Closure Properties pane and enable time dependence for the output contact closure. You can specify the on/off timing of the contact closure signal sent to an external device. The area to the right of the Output Contact Closure icon graphically displays the on/off status of the contact closure over time as a timeline.
In the table, you can import or export settings using a .csv, .txt, or .xml file.
NOTE
You must configure the Input Contact Closure setting in the Thermo Foundation Instrument Configuration.
The following table describes the parameters in the Output Contact Closure Properties pane.
Parameter | Description |
|---|---|
Time (min) | The time (in minutes) when the contact closure state changes. Range: 0 to the maximum method duration (5,000 maximum) |
Position | The contact closure signal is Open or Closed. Default: Open |