Select the Divert Valve checkbox to open the Divert Valve Properties pane and enable time dependence for the divert valve. The area to the right of the Divert Valve icon graphically displays the time dependence of the divert valve position as a timeline.
In the divert valve position table, you can import or export settings using a .csv, .txt, or .xml file.
NOTE
You can configure Divert Valve A and or Divert Valve B depending your instrument configuration.
The following table describes the parameters in the Divert Valve Properties pane.
Parameter | Description |
|---|---|
Time (min) | The time (in minutes) when the divert valve changes position. Range: 0 to the maximum method duration (5,000 maximum) |
Position | The divert valve position: 1-2 (default) or 1-6. |