After you create an experiment workflow, save your changes as an instrument method.
Procedure
  1. From the top menu bar, select File → Save As.
  2. Specify a location and name for your instrument method, and then select Save.
NOTE
The instrument method file has a .meth file extension.