Use the Method Timeline area to create a timeline for your experiment. Each timeline is a graphical representation of the experiment’s specified time.
The following table describes the parameters in the Method Timeline area.
Parameter | Description |
|---|---|
Method Duration (min) | Specify the time duration (in minutes) for the instrument method. The method duration area displays the remaining experiment time (in minutes). Range: 0.1–5,000; default: 60 |
Zoom control buttons | The zoom control buttons control the timeline display. From top to bottom:
|