Use the Sort by m/z filter to set the sorting order of the precursor ions by using their m/z values.
NOTE
To use the "Sort by m/z" and "Sort by Charge" filters in the same scan branch, you must add the "Sort by Charge" filter first.
The following table describes the parameters in the Sort by m/z Properties pane.
Parameter | Description |
|---|---|
Precursor Priority | Select from the following options:
|