Use the Sort by Intensity filter to set the sorting order of the precursor ions using their intensity values.
NOTE
To use the "Sort by Intensity" and "Sort by Charge" filters in the same scan branch, you must add the "Sort by Charge" filter first.
The following table describes the parameters in the Sort by Intensity Properties pane.
Parameter | Description |
|---|---|
Precursor Priority | Select from the following options:
|