The Targeted Loss Ion Exclusion filter prevents the system from performing a data-dependent scan if it detects a targeted loss from the list. If the MSn level of the appended data-dependent scan is the same as the previous scan, the system performs data-dependent analysis of the same precursor. If the MSn level of the appended data-dependent scan is n + 1, the system performs data-dependent analysis on the top N most intense ions. You can also import the settings of the mass list table from (or export them to) a .cvs, a .txt, or an .xml file.
The following table describes the parameters in the Targeted Loss Exclusion Properties pane.
Parameter | Description |
|---|---|
Mass List Type | Select from the following mass list types:
|
| |
Parameters in the Mass List Table | |
Compound | (Optional) Specify the name of the ion. |
m/z | (Available when the Mass List Type is set to "m/z".) Specify the m/z of the ion on which the MS performs the scan. When Formula is defined, the m/z is calculated automatically. The accepted m/z range will vary depending on analyzer used (Quadrupole or Ion Trap) and HMRn+ license activation. |
Formula | (Available when the Mass List Type is set to "M".) (Optional) Define the compound formula (for example, CH3OH) or peptide sequence (for example, MRFA). |
M | (Available when the Mass List Type is set to "M".) The targeted loss mass (in Daltons). Range: 0–100,000; default: 523.2577 |
z loss | (Available when the Mass List Type is set to "M".) Optional, in some cases. Specify the expected charge state of the precursor ion, which is used to determine collision energy or reaction times (when applicable). When 'Formula' is defined, specify the expected charge state of the precursor ion. This is used to calculate the m/z. Range: 0–24; default: 1 |
| |
Exclusion mass width | Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
|
Low | The low side of the mass tolerance width window. The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
High | The high side of the mass tolerance width window. The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
Trigger Only with Detection of Correct Charge State of the Product Ion | (Available when the Mass List Type is set to "M".) Performs subsequent scans only if the product ion charge state exactly matches the targeted ion charge state. Default: Selected |
Ignore Charge State Requirement for Unassigned Ions | (Available when the Mass List Type is set to "M".) When selected, ions that have an unassigned charge state do not need to meet the charge-state requirement in the table. Default: Selected |