The Precursor Ion Exclusion filter excludes ions from triggering a data-dependent scan if they are inside the mass tolerance window of the precursor ion.
The following table describes the parameters in the Precursor Ion Exclusion Properties pane.
Parameter
Description
Exclusion mass width
Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
  • ppm: The low and high values of the exclusion mass width in parts per million (ppm) of the reference mass. This option is appropriate for the Orbitrap mass analyzer because the peak width is proportional to the m/z.
  • You can specify the exclusion mass width as a percent value of the reference mass:
    • First select the ppm option, and then type the desired values in the Low box and High box. The exclusion mass is expressed in parts per million (ppm). When you select ppm, the size of the window around the reference mass is proportional to the m/z of the reference mass.
  • m/z: The low and high values of the exclusion mass width in m/z. This option is appropriate for the ion trap mass analyzer because the peak width is constant.
  • You can specify the exclusion mass width in m/z:
    • First select the m/z option, and then type the desired values in the Low box and High box. When you select m/z, the size of the window around the reference mass remains the same regardless of the m/z of the reference mass.
Low
The low side of the mass tolerance width window.
The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak.
Range (ppm): 0.01–1,000,000; default: 25
Range (m/z): 0.01–100; default: 0.5
High
The high side of the mass tolerance width window.
The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak.
Range (ppm): 0.01–1,000,000; default: 25
Range (m/z): 0.01–100; default: 0.5